Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160680899-160681172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD48
Entrez Gene ID
962
Gene Name
CD48 molecule
Gene Aliases
BCM1, BLAST, BLAST1, MEM-102, SLAMF2, hCD48, mCD48
UniGene
Hs.243564
Gene Chromosome Location
Chr.1: 160678746-160711851 on Build GRCh38
Transcripts
NM_001256030.1 NM_001778.3 XM_005245625.1 XM_011510171.1 XM_017002867.1

SNP Detail

SNP ID Chromosome Location
rs754715652 Chr.1: 160681050-160681050 on Build GRCh38
rs190382630 Chr.1: 160681129-160681129 on Build GRCh38
rs561529017 Chr.1: 160681013-160681013 on Build GRCh38
rs958994512 Chr.1: 160681069-160681069 on Build GRCh38
rs916237586 Chr.1: 160680975-160680975 on Build GRCh38
rs181648829 Chr.1: 160681105-160681105 on Build GRCh38
rs4656926 Chr.1: 160681123-160681123 on Build GRCh38
rs778535449 Chr.1: 160681131-160681131 on Build GRCh38
rs3835477 Chr.1: 160681003-160681003 on Build GRCh38
rs532842428 Chr.1: 160681097-160681097 on Build GRCh38
rs767361486 Chr.1: 160681121-160681121 on Build GRCh38
rs142100077 Chr.1: 160681129-160681129 on Build GRCh38
rs6667145 Chr.1: 160681148-160681148 on Build GRCh38
rs569477186 Chr.1: 160681114-160681114 on Build GRCh38
rs771255456 Chr.1: 160681101-160681101 on Build GRCh38
rs549215035 Chr.1: 160681125-160681125 on Build GRCh38
rs751823707 Chr.1: 160681112-160681112 on Build GRCh38
rs543272728 Chr.1: 160680977-160680977 on Build GRCh38
rs536506546 Chr.1: 160681028-160681028 on Build GRCh38
rs114879544 Chr.1: 160681063-160681063 on Build GRCh38
rs991759707 Chr.1: 160681079-160681079 on Build GRCh38
rs563853525 Chr.1: 160680939-160680939 on Build GRCh38
rs908591950 Chr.1: 160680964-160680964 on Build GRCh38
rs776348333 Chr.1: 160681127-160681128 on Build GRCh38
rs990758883 Chr.1: 160680971-160680971 on Build GRCh38
rs1031995916 Chr.1: 160680921-160680921 on Build GRCh38
rs770648800 Chr.1: 160681068-160681068 on Build GRCh38
rs373252598 Chr.1: 160680940-160680940 on Build GRCh38
rs796214715 Chr.1: 160681013-160681014 on Build GRCh38
rs957631669 Chr.1: 160680929-160680929 on Build GRCh38
rs1051675500 Chr.1: 160681048-160681048 on Build GRCh38
rs535058549 Chr.1: 160681130-160681130 on Build GRCh38
rs748153951 Chr.1: 160681060-160681060 on Build GRCh38
rs745582708 Chr.1: 160681132-160681132 on Build GRCh38
rs771834935 Chr.1: 160681136-160681136 on Build GRCh38
rs780938768 Chr.1: 160681053-160681053 on Build GRCh38
rs992145645 Chr.1: 160680933-160680933 on Build GRCh38
rs766641338 Chr.1: 160681111-160681111 on Build GRCh38
rs574409891 Chr.1: 160680963-160680963 on Build GRCh38
rs981571215 Chr.1: 160680947-160680947 on Build GRCh38
rs745492862 Chr.1: 160681084-160681084 on Build GRCh38
rs765603144 Chr.1: 160681103-160681103 on Build GRCh38
rs898500889 Chr.1: 160680989-160680989 on Build GRCh38
rs971604907 Chr.1: 160681092-160681092 on Build GRCh38
rs1020713133 Chr.1: 160681062-160681062 on Build GRCh38
rs1050072548 Chr.1: 160681017-160681017 on Build GRCh38
rs912751189 Chr.1: 160681052-160681052 on Build GRCh38
rs188508527 Chr.1: 160681014-160681014 on Build GRCh38
rs781699211 Chr.1: 160681141-160681141 on Build GRCh38
rs960590252 Chr.1: 160680942-160680942 on Build GRCh38
rs397841059 Chr.1: 160681007-160681007 on Build GRCh38
rs748681739 Chr.1: 160681065-160681065 on Build GRCh38
rs932602843 Chr.1: 160681009-160681009 on Build GRCh38
rs749645176 Chr.1: 160681064-160681064 on Build GRCh38
rs916091302 Chr.1: 160680934-160680934 on Build GRCh38
rs1038359251 Chr.1: 160680984-160680984 on Build GRCh38
rs769181478 Chr.1: 160681085-160681085 on Build GRCh38
rs547065869 Chr.1: 160681029-160681029 on Build GRCh38
rs908698754 Chr.1: 160681113-160681113 on Build GRCh38
rs759754138 Chr.1: 160681115-160681115 on Build GRCh38
rs774098485 Chr.1: 160681075-160681075 on Build GRCh38

View Full Product Details