Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55651220-55651453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC106
Entrez Gene ID
29903
Gene Name
coiled-coil domain containing 106
Gene Aliases
HSU79303, ZNF581
UniGene
Hs.82482
Gene Chromosome Location
Chr.19: 55647588-55653161 on Build GRCh38
Transcripts
NM_013301.2 XM_005258827.2 XM_005258828.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390823 c.453G>A p.R151R Chr.19: 55651422-55651422 on Build GRCh38
6205612 c.382G>A p.E128K Chr.19: 55651351-55651351 on Build GRCh38
6387756 c.323G>A p.C108Y Chr.19: 55651292-55651292 on Build GRCh38
1001587 c.391G>T p.D131Y Chr.19: 55651360-55651360 on Build GRCh38
7286844 c.381C>T p.G127G Chr.19: 55651350-55651350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983810277 Chr.19: 55651265-55651265 on Build GRCh38
rs772304153 Chr.19: 55651284-55651284 on Build GRCh38
rs2287791 Chr.19: 55651410-55651410 on Build GRCh38
rs556667212 Chr.19: 55651393-55651393 on Build GRCh38
rs147212550 Chr.19: 55651391-55651391 on Build GRCh38
rs201475292 Chr.19: 55651273-55651273 on Build GRCh38
rs771966660 Chr.19: 55651422-55651422 on Build GRCh38
rs756318484 Chr.19: 55651258-55651258 on Build GRCh38
rs1042482109 Chr.19: 55651397-55651397 on Build GRCh38
rs2459750 Chr.19: 55651332-55651332 on Build GRCh38
rs138679001 Chr.19: 55651398-55651398 on Build GRCh38
rs760815871 Chr.19: 55651336-55651336 on Build GRCh38
rs200120422 Chr.19: 55651399-55651399 on Build GRCh38
rs201340136 Chr.19: 55651390-55651390 on Build GRCh38
rs764656310 Chr.19: 55651260-55651263 on Build GRCh38
rs766746051 Chr.19: 55651342-55651342 on Build GRCh38
rs781485981 Chr.19: 55651402-55651402 on Build GRCh38
rs748969572 Chr.19: 55651404-55651404 on Build GRCh38
rs200684341 Chr.19: 55651414-55651414 on Build GRCh38
rs779679255 Chr.19: 55651359-55651359 on Build GRCh38
rs774132352 Chr.19: 55651324-55651324 on Build GRCh38
rs758903377 Chr.19: 55651350-55651350 on Build GRCh38
rs374434219 Chr.19: 55651423-55651423 on Build GRCh38
rs535595597 Chr.19: 55651337-55651337 on Build GRCh38
rs61736518 Chr.19: 55651346-55651346 on Build GRCh38
rs771983589 Chr.19: 55651383-55651383 on Build GRCh38
rs373493546 Chr.19: 55651333-55651333 on Build GRCh38
rs750165054 Chr.19: 55651334-55651334 on Build GRCh38
rs1051075 Chr.19: 55651429-55651429 on Build GRCh38
rs534270066 Chr.19: 55651261-55651261 on Build GRCh38
rs755598237 Chr.19: 55651269-55651269 on Build GRCh38
rs779615835 Chr.19: 55651344-55651344 on Build GRCh38
rs748589063 Chr.19: 55651274-55651274 on Build GRCh38
rs568364976 Chr.19: 55651242-55651242 on Build GRCh38
rs761660396 Chr.19: 55651381-55651381 on Build GRCh38
rs554247732 Chr.19: 55651343-55651343 on Build GRCh38
rs769496373 Chr.19: 55651358-55651358 on Build GRCh38
rs760540069 Chr.19: 55651387-55651387 on Build GRCh38
rs867449354 Chr.19: 55651405-55651405 on Build GRCh38
rs150361758 Chr.19: 55651316-55651316 on Build GRCh38
rs752759766 Chr.19: 55651394-55651394 on Build GRCh38
rs754558291 Chr.19: 55651408-55651408 on Build GRCh38
rs200256103 Chr.19: 55651395-55651395 on Build GRCh38
rs577202323 Chr.19: 55651323-55651323 on Build GRCh38
rs371292659 Chr.19: 55651315-55651315 on Build GRCh38
rs773167474 Chr.19: 55651427-55651427 on Build GRCh38
rs751609209 Chr.19: 55651397-55651411 on Build GRCh38
rs998389992 Chr.19: 55651239-55651239 on Build GRCh38
rs1015860468 Chr.19: 55651389-55651389 on Build GRCh38
rs773871129 Chr.19: 55651380-55651380 on Build GRCh38
rs574675982 Chr.19: 55651415-55651415 on Build GRCh38
rs745541102 Chr.19: 55651294-55651294 on Build GRCh38
rs748844748 Chr.19: 55651365-55651365 on Build GRCh38
rs143763452 Chr.19: 55651378-55651378 on Build GRCh38
rs778154930 Chr.19: 55651289-55651289 on Build GRCh38
rs989304394 Chr.19: 55651320-55651320 on Build GRCh38
rs536663437 Chr.19: 55651250-55651250 on Build GRCh38
rs894582413 Chr.19: 55651357-55651357 on Build GRCh38
rs769370058 Chr.19: 55651309-55651309 on Build GRCh38

View Full Product Details