Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73737316-73737541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD11
Entrez Gene ID
83451
Gene Name
abhydrolase domain containing 11
Gene Aliases
PP1226, WBSCR21
UniGene
Hs.647045
Gene Chromosome Location
Chr.7: 73736094-73738867 on Build GRCh38
Transcripts
NM_001145364.2 NM_001301058.1 NM_001321382.1 NM_001321383.1 NM_148912.3 NM_148913.3 NR_026912.2 NR_135627.1 XM_006716144.2 XM_011516590.2 XM_011516591.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4770822 c.503C>T p.P168L Chr.7: 73737351-73737351 on Build GRCh38
6438395 c.525_526CC>TT p.H176Y Chr.7: 73737328-73737329 on Build GRCh38
4771682 c.481C>G p.L161V Chr.7: 73737373-73737373 on Build GRCh38
3698534 c.531T>A p.F177L Chr.7: 73737323-73737323 on Build GRCh38
4187251 c.511delA p.S171fs*14 Chr.7: 73737343-73737343 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782036813 Chr.7: 73737511-73737511 on Build GRCh38
rs782814264 Chr.7: 73737358-73737358 on Build GRCh38
rs781821890 Chr.7: 73737380-73737380 on Build GRCh38
rs782608700 Chr.7: 73737440-73737440 on Build GRCh38
rs370448641 Chr.7: 73737406-73737406 on Build GRCh38
rs141117786 Chr.7: 73737433-73737433 on Build GRCh38
rs782116973 Chr.7: 73737354-73737354 on Build GRCh38
rs782018129 Chr.7: 73737360-73737360 on Build GRCh38
rs994226465 Chr.7: 73737352-73737352 on Build GRCh38
rs782742433 Chr.7: 73737365-73737365 on Build GRCh38
rs782263005 Chr.7: 73737441-73737441 on Build GRCh38
rs782542113 Chr.7: 73737414-73737414 on Build GRCh38
rs782068817 Chr.7: 73737375-73737375 on Build GRCh38
rs782275398 Chr.7: 73737516-73737516 on Build GRCh38
rs147960131 Chr.7: 73737412-73737412 on Build GRCh38
rs200897838 Chr.7: 73737376-73737376 on Build GRCh38
rs782375030 Chr.7: 73737443-73737443 on Build GRCh38
rs782773522 Chr.7: 73737407-73737407 on Build GRCh38
rs782496232 Chr.7: 73737438-73737438 on Build GRCh38
rs781801229 Chr.7: 73737498-73737498 on Build GRCh38

View Full Product Details