Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218960519-218960792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5R2
Entrez Gene ID
8941
Gene Name
cyclin dependent kinase 5 regulatory subunit 2
Gene Aliases
NCK5AI, P39, p39nck5ai
UniGene
Hs.158460
Gene Chromosome Location
Chr.2: 218959628-218962155 on Build GRCh38
Transcripts
NM_003936.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406474 c.865G>A p.D289N Chr.2: 218960685-218960685 on Build GRCh38
3044189 c.768C>G p.A256A Chr.2: 218960588-218960588 on Build GRCh38
4393333 c.729C>T p.C243C Chr.2: 218960549-218960549 on Build GRCh38
6625541 c.762G>A p.S254S Chr.2: 218960582-218960582 on Build GRCh38
3044197 c.960G>A p.Q320Q Chr.2: 218960780-218960780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758578518 Chr.2: 218960716-218960716 on Build GRCh38
rs757381329 Chr.2: 218960765-218960765 on Build GRCh38
rs370057429 Chr.2: 218960727-218960727 on Build GRCh38
rs533673689 Chr.2: 218960558-218960558 on Build GRCh38
rs772855228 Chr.2: 218960622-218960622 on Build GRCh38
rs779904283 Chr.2: 218960563-218960563 on Build GRCh38
rs768379674 Chr.2: 218960600-218960600 on Build GRCh38
rs747780319 Chr.2: 218960610-218960610 on Build GRCh38
rs765570302 Chr.2: 218960673-218960673 on Build GRCh38
rs996683804 Chr.2: 218960738-218960738 on Build GRCh38
rs758622919 Chr.2: 218960555-218960555 on Build GRCh38
rs749207583 Chr.2: 218960587-218960587 on Build GRCh38
rs747047996 Chr.2: 218960559-218960559 on Build GRCh38
rs777345085 Chr.2: 218960648-218960648 on Build GRCh38
rs765629753 Chr.2: 218960540-218960540 on Build GRCh38
rs760088475 Chr.2: 218960660-218960660 on Build GRCh38
rs866602801 Chr.2: 218960654-218960654 on Build GRCh38
rs753054357 Chr.2: 218960695-218960695 on Build GRCh38
rs950647365 Chr.2: 218960682-218960682 on Build GRCh38
rs771718118 Chr.2: 218960621-218960621 on Build GRCh38
rs765974378 Chr.2: 218960631-218960631 on Build GRCh38
rs751649795 Chr.2: 218960749-218960749 on Build GRCh38
rs756047603 Chr.2: 218960562-218960562 on Build GRCh38
rs774178690 Chr.2: 218960601-218960601 on Build GRCh38

View Full Product Details