Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92343029-92343266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKIB1
Entrez Gene ID
54467
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
-
UniGene
Hs.83293
Gene Chromosome Location
Chr.7: 92246216-92401773 on Build GRCh38
Transcripts
NM_019004.1 XM_005250458.3 XM_011516334.1 XM_011516335.2 XM_017012359.1 XM_017012360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4510928 c.851C>T p.S284L Chr.7: 92343087-92343087 on Build GRCh38
1182818 c.871C>T p.P291S Chr.7: 92343107-92343107 on Build GRCh38
5584447 c.862A>C p.M288L Chr.7: 92343098-92343098 on Build GRCh38
7169112 c.958G>A p.E320K Chr.7: 92343194-92343194 on Build GRCh38
1623302 c.916A>G p.R306G Chr.7: 92343152-92343152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749192724 Chr.7: 92343139-92343139 on Build GRCh38
rs758831784 Chr.7: 92343067-92343067 on Build GRCh38
rs747778228 Chr.7: 92343116-92343116 on Build GRCh38
rs751054587 Chr.7: 92343104-92343104 on Build GRCh38
rs754461918 Chr.7: 92343228-92343228 on Build GRCh38
rs991054022 Chr.7: 92343126-92343126 on Build GRCh38
rs765539705 Chr.7: 92343063-92343063 on Build GRCh38
rs768909638 Chr.7: 92343172-92343172 on Build GRCh38
rs780791692 Chr.7: 92343112-92343112 on Build GRCh38
rs774499835 Chr.7: 92343142-92343142 on Build GRCh38
rs747057661 Chr.7: 92343170-92343170 on Build GRCh38
rs983591478 Chr.7: 92343225-92343225 on Build GRCh38
rs767044991 Chr.7: 92343232-92343232 on Build GRCh38
rs752100081 Chr.7: 92343227-92343227 on Build GRCh38
rs763431028 Chr.7: 92343207-92343207 on Build GRCh38
rs762211381 Chr.7: 92343060-92343060 on Build GRCh38
rs765815046 Chr.7: 92343097-92343097 on Build GRCh38
rs761968945 Chr.7: 92343179-92343179 on Build GRCh38
rs766865093 Chr.7: 92343220-92343220 on Build GRCh38
rs755899387 Chr.7: 92343125-92343125 on Build GRCh38
rs765567969 Chr.7: 92343187-92343187 on Build GRCh38
rs777436339 Chr.7: 92343138-92343138 on Build GRCh38
rs750899321 Chr.7: 92343064-92343064 on Build GRCh38
rs768661776 Chr.7: 92343056-92343056 on Build GRCh38

View Full Product Details