Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107744033-107744304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLL1
Entrez Gene ID
79872
Gene Name
Cbl proto-oncogene like 1
Gene Aliases
HAKAI, RNF188
UniGene
Hs.592271
Gene Chromosome Location
Chr.7: 107743497-107764997 on Build GRCh38
Transcripts
NM_001284291.1 NM_024814.3 NR_024199.2 XM_011516580.2 XM_017012643.1 XM_017012644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432112 c.7C>T p.H3Y Chr.7: 107744170-107744170 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139250805 Chr.7: 107744092-107744092 on Build GRCh38
rs916732193 Chr.7: 107744195-107744195 on Build GRCh38
rs1017843025 Chr.7: 107744214-107744214 on Build GRCh38
rs935014216 Chr.7: 107744256-107744256 on Build GRCh38
rs750571801 Chr.7: 107744228-107744228 on Build GRCh38
rs765469996 Chr.7: 107744227-107744227 on Build GRCh38
rs1015029392 Chr.7: 107744122-107744122 on Build GRCh38
rs371231337 Chr.7: 107744068-107744068 on Build GRCh38
rs751404862 Chr.7: 107744124-107744124 on Build GRCh38
rs1010712012 Chr.7: 107744062-107744062 on Build GRCh38
rs1039532931 Chr.7: 107744268-107744268 on Build GRCh38
rs199725875 Chr.7: 107744224-107744224 on Build GRCh38
rs762074182 Chr.7: 107744217-107744217 on Build GRCh38
rs769073308 Chr.7: 107744206-107744206 on Build GRCh38
rs964924934 Chr.7: 107744216-107744216 on Build GRCh38
rs1044356982 Chr.7: 107744084-107744084 on Build GRCh38
rs777187340 Chr.7: 107744158-107744158 on Build GRCh38
rs747866289 Chr.7: 107744147-107744147 on Build GRCh38
rs893506781 Chr.7: 107744058-107744058 on Build GRCh38
rs983307679 Chr.7: 107744075-107744075 on Build GRCh38
rs935713334 Chr.7: 107744118-107744118 on Build GRCh38
rs1005254884 Chr.7: 107744104-107744104 on Build GRCh38
rs1030786226 Chr.7: 107744071-107744071 on Build GRCh38
rs780072684 Chr.7: 107744119-107744119 on Build GRCh38
rs375148180 Chr.7: 107744127-107744127 on Build GRCh38
rs369676926 Chr.7: 107744213-107744213 on Build GRCh38
rs547129711 Chr.7: 107744263-107744263 on Build GRCh38
rs769546676 Chr.7: 107744157-107744157 on Build GRCh38
rs761101274 Chr.7: 107744204-107744204 on Build GRCh38
rs530500617 Chr.7: 107744066-107744066 on Build GRCh38
rs973599521 Chr.7: 107744172-107744172 on Build GRCh38
rs946455597 Chr.7: 107744266-107744266 on Build GRCh38
rs748860707 Chr.7: 107744175-107744175 on Build GRCh38
rs564766694 Chr.7: 107744194-107744194 on Build GRCh38
rs890603113 Chr.7: 107744185-107744185 on Build GRCh38
rs950838632 Chr.7: 107744279-107744279 on Build GRCh38
rs920725495 Chr.7: 107744053-107744053 on Build GRCh38
rs928173669 Chr.7: 107744235-107744235 on Build GRCh38
rs998037841 Chr.7: 107744070-107744070 on Build GRCh38
rs369561628 Chr.7: 107744069-107744069 on Build GRCh38
rs1052506708 Chr.7: 107744257-107744257 on Build GRCh38
rs1039484012 Chr.7: 107744065-107744065 on Build GRCh38
rs949966439 Chr.7: 107744082-107744082 on Build GRCh38
rs561689517 Chr.7: 107744060-107744060 on Build GRCh38
rs781068267 Chr.7: 107744136-107744136 on Build GRCh38
rs770294559 Chr.7: 107744183-107744183 on Build GRCh38
rs372259387 Chr.7: 107744105-107744105 on Build GRCh38
rs917058248 Chr.7: 107744081-107744081 on Build GRCh38
rs373998247 Chr.7: 107744182-107744182 on Build GRCh38
rs73417978 Chr.7: 107744280-107744280 on Build GRCh38
rs915004726 Chr.7: 107744258-107744258 on Build GRCh38
rs963430217 Chr.7: 107744150-107744150 on Build GRCh38

View Full Product Details