Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12553228-12553495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1D
Entrez Gene ID
57118
Gene Name
calcium/calmodulin dependent protein kinase ID
Gene Aliases
CKLiK, CaM-K1, CaMKID
UniGene
Hs.600547
Gene Chromosome Location
Chr.10: 12349514-12835545 on Build GRCh38
Transcripts
NM_020397.3 NM_153498.3 XM_006717481.3 XM_006717482.3 XM_006717483.3 XM_011519591.2 XM_017016438.1 XM_017016439.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3435428 c.99C>T p.A33A Chr.10: 12553231-12553231 on Build GRCh38
465393 c.219G>T p.L73L Chr.10: 12553351-12553351 on Build GRCh38
6436413 c.149C>A p.A50D Chr.10: 12553281-12553281 on Build GRCh38
6436412 c.149C>A p.A50D Chr.10: 12553281-12553281 on Build GRCh38
465394 c.219G>T p.L73L Chr.10: 12553351-12553351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753517881 Chr.10: 12553299-12553299 on Build GRCh38
rs553333737 Chr.10: 12553306-12553306 on Build GRCh38
rs932962488 Chr.10: 12553438-12553438 on Build GRCh38
rs765910028 Chr.10: 12553370-12553370 on Build GRCh38
rs767447307 Chr.10: 12553356-12553356 on Build GRCh38
rs1039387914 Chr.10: 12553421-12553421 on Build GRCh38
rs368170335 Chr.10: 12553374-12553374 on Build GRCh38
rs757633702 Chr.10: 12553380-12553380 on Build GRCh38
rs139664294 Chr.10: 12553457-12553457 on Build GRCh38
rs767392318 Chr.10: 12553261-12553261 on Build GRCh38
rs760162495 Chr.10: 12553365-12553365 on Build GRCh38
rs769076898 Chr.10: 12553400-12553400 on Build GRCh38
rs751503100 Chr.10: 12553458-12553458 on Build GRCh38
rs768486499 Chr.10: 12553346-12553346 on Build GRCh38
rs909074433 Chr.10: 12553382-12553382 on Build GRCh38
rs749966956 Chr.10: 12553269-12553269 on Build GRCh38
rs751423327 Chr.10: 12553445-12553445 on Build GRCh38
rs745945368 Chr.10: 12553315-12553315 on Build GRCh38
rs758758416 Chr.10: 12553305-12553305 on Build GRCh38
rs544885345 Chr.10: 12553461-12553461 on Build GRCh38
rs890555350 Chr.10: 12553470-12553470 on Build GRCh38
rs780157660 Chr.10: 12553404-12553404 on Build GRCh38
rs571244811 Chr.10: 12553345-12553345 on Build GRCh38
rs1031738398 Chr.10: 12553467-12553467 on Build GRCh38
rs41306818 Chr.10: 12553403-12553403 on Build GRCh38
rs55749722 Chr.10: 12553312-12553312 on Build GRCh38
rs370966780 Chr.10: 12553253-12553253 on Build GRCh38
rs980794056 Chr.10: 12553302-12553302 on Build GRCh38
rs573595156 Chr.10: 12553429-12553429 on Build GRCh38
rs1077745 Chr.10: 12553428-12553428 on Build GRCh38
rs773076177 Chr.10: 12553359-12553359 on Build GRCh38
rs56079552 Chr.10: 12553351-12553351 on Build GRCh38
rs781753117 Chr.10: 12553314-12553314 on Build GRCh38
rs114093778 Chr.10: 12553376-12553376 on Build GRCh38
rs537697759 Chr.10: 12553294-12553294 on Build GRCh38
rs757864847 Chr.10: 12553307-12553307 on Build GRCh38
rs765962732 Chr.10: 12553297-12553297 on Build GRCh38
rs769900753 Chr.10: 12553322-12553322 on Build GRCh38
rs759272395 Chr.10: 12553379-12553379 on Build GRCh38
rs983158466 Chr.10: 12553387-12553387 on Build GRCh38
rs371289261 Chr.10: 12553375-12553375 on Build GRCh38
rs1021785288 Chr.10: 12553471-12553471 on Build GRCh38
rs781778179 Chr.10: 12553402-12553402 on Build GRCh38
rs34194224 Chr.10: 12553330-12553330 on Build GRCh38
rs199673286 Chr.10: 12553401-12553401 on Build GRCh38
rs1077744 Chr.10: 12553444-12553444 on Build GRCh38
rs761555171 Chr.10: 12553355-12553355 on Build GRCh38
rs774085044 Chr.10: 12553403-12553404 on Build GRCh38
rs972745162 Chr.10: 12553332-12553332 on Build GRCh38
rs946623103 Chr.10: 12553448-12553448 on Build GRCh38
rs775427069 Chr.10: 12553406-12553406 on Build GRCh38
rs891509053 Chr.10: 12553449-12553449 on Build GRCh38

View Full Product Details