Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116389366-116389637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A1
Entrez Gene ID
476
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Gene Aliases
-
UniGene
Hs.371889
Gene Chromosome Location
Chr.1: 116373173-116404774 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3862330 c.903C>T p.F301F Chr.1: 116389587-116389587 on Build GRCh38
3471873 c.937C>T p.L313F Chr.1: 116389621-116389621 on Build GRCh38
4654662 c.877C>T p.H293Y Chr.1: 116389561-116389561 on Build GRCh38
5843180 c.817C>T p.L273F Chr.1: 116389501-116389501 on Build GRCh38
4664263 c.802G>T p.G268* Chr.1: 116389486-116389486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912110487 Chr.1: 116389467-116389467 on Build GRCh38
rs748460069 Chr.1: 116389410-116389410 on Build GRCh38
rs866032260 Chr.1: 116389514-116389514 on Build GRCh38
rs200841398 Chr.1: 116389595-116389595 on Build GRCh38
rs749677403 Chr.1: 116389403-116389403 on Build GRCh38
rs564117347 Chr.1: 116389470-116389470 on Build GRCh38
rs774561525 Chr.1: 116389409-116389409 on Build GRCh38
rs930844243 Chr.1: 116389607-116389607 on Build GRCh38
rs752531768 Chr.1: 116389517-116389517 on Build GRCh38
rs760646881 Chr.1: 116389430-116389430 on Build GRCh38
rs866444285 Chr.1: 116389573-116389573 on Build GRCh38
rs776666372 Chr.1: 116389443-116389443 on Build GRCh38
rs780584299 Chr.1: 116389526-116389527 on Build GRCh38
rs765069962 Chr.1: 116389475-116389475 on Build GRCh38
rs768969158 Chr.1: 116389404-116389404 on Build GRCh38
rs1036506727 Chr.1: 116389463-116389463 on Build GRCh38
rs763626611 Chr.1: 116389581-116389581 on Build GRCh38
rs966706568 Chr.1: 116389509-116389509 on Build GRCh38
rs772179315 Chr.1: 116389414-116389414 on Build GRCh38
rs940435898 Chr.1: 116389436-116389436 on Build GRCh38
rs773380582 Chr.1: 116389424-116389424 on Build GRCh38
rs77346051 Chr.1: 116389583-116389583 on Build GRCh38
rs780517789 Chr.1: 116389389-116389389 on Build GRCh38
rs759405523 Chr.1: 116389447-116389447 on Build GRCh38
rs899341780 Chr.1: 116389518-116389518 on Build GRCh38
rs758139508 Chr.1: 116389530-116389530 on Build GRCh38
rs371630565 Chr.1: 116389434-116389434 on Build GRCh38

View Full Product Details