Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 65077777-65078050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP68
Entrez Gene ID
23177
Gene Name
centrosomal protein 68
Gene Aliases
KIAA0582
UniGene
Hs.709257
Gene Chromosome Location
Chr.2: 65056361-65087008 on Build GRCh38
Transcripts
NM_001319100.1 NM_001319101.1 NM_015147.2 NR_134966.1 XM_005264219.3 XM_017003633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1248103 c.2037G>C p.Q679H Chr.2: 65077897-65077897 on Build GRCh38
3055262 c.2068G>A p.E690K Chr.2: 65077928-65077928 on Build GRCh38
5059003 c.2082G>T p.Q694H Chr.2: 65077942-65077942 on Build GRCh38
6422784 c.2102C>A p.P701Q Chr.2: 65077962-65077962 on Build GRCh38
4927451 c.2039C>G p.S680C Chr.2: 65077899-65077899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776481678 Chr.2: 65078016-65078016 on Build GRCh38
rs377568170 Chr.2: 65077830-65077830 on Build GRCh38
rs1052759617 Chr.2: 65077806-65077806 on Build GRCh38
rs576833128 Chr.2: 65077905-65077905 on Build GRCh38
rs760041664 Chr.2: 65077906-65077906 on Build GRCh38
rs748048178 Chr.2: 65077853-65077853 on Build GRCh38
rs770698622 Chr.2: 65078007-65078007 on Build GRCh38
rs373721627 Chr.2: 65077859-65077859 on Build GRCh38
rs989959238 Chr.2: 65078029-65078029 on Build GRCh38
rs754862392 Chr.2: 65077988-65077988 on Build GRCh38
rs114452676 Chr.2: 65077826-65077826 on Build GRCh38
rs766781808 Chr.2: 65077991-65077992 on Build GRCh38
rs373907873 Chr.2: 65077994-65077994 on Build GRCh38
rs762806146 Chr.2: 65077857-65077858 on Build GRCh38
rs367720438 Chr.2: 65077997-65077997 on Build GRCh38
rs749130283 Chr.2: 65077837-65077837 on Build GRCh38
rs764794767 Chr.2: 65077811-65077811 on Build GRCh38
rs746552835 Chr.2: 65078004-65078004 on Build GRCh38
rs779777161 Chr.2: 65077831-65077831 on Build GRCh38
rs751664315 Chr.2: 65077958-65077958 on Build GRCh38
rs371059807 Chr.2: 65077838-65077838 on Build GRCh38
rs770893967 Chr.2: 65077951-65077953 on Build GRCh38
rs753627978 Chr.2: 65077977-65077977 on Build GRCh38
rs143036273 Chr.2: 65077912-65077912 on Build GRCh38
rs757450495 Chr.2: 65077962-65077962 on Build GRCh38
rs369695066 Chr.2: 65077999-65077999 on Build GRCh38
rs971759150 Chr.2: 65077972-65077972 on Build GRCh38
rs994610846 Chr.2: 65077911-65077911 on Build GRCh38
rs957210463 Chr.2: 65078006-65078006 on Build GRCh38
rs553603477 Chr.2: 65077870-65077870 on Build GRCh38
rs768552056 Chr.2: 65077840-65077840 on Build GRCh38
rs760354181 Chr.2: 65077880-65077880 on Build GRCh38
rs376693641 Chr.2: 65077935-65077935 on Build GRCh38
rs777314861 Chr.2: 65078003-65078003 on Build GRCh38
rs905815084 Chr.2: 65077910-65077910 on Build GRCh38
rs766180522 Chr.2: 65077883-65077883 on Build GRCh38
rs778575579 Chr.2: 65077995-65077995 on Build GRCh38
rs765646512 Chr.2: 65077908-65077908 on Build GRCh38
rs777730454 Chr.2: 65077823-65077823 on Build GRCh38
rs370498293 Chr.2: 65077974-65077974 on Build GRCh38
rs200409745 Chr.2: 65077860-65077860 on Build GRCh38
rs774415330 Chr.2: 65077842-65077842 on Build GRCh38

View Full Product Details