Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89612201-89612456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD6
Entrez Gene ID
22881
Gene Name
ankyrin repeat domain 6
Gene Aliases
-
UniGene
Hs.702213
Gene Chromosome Location
Chr.6: 89433178-89633834 on Build GRCh38
Transcripts
NM_001242809.1 NM_001242811.1 NM_001242813.1 NM_001242814.1 NM_014942.4 XM_005248679.3 XM_005248680.3 XM_005248681.2 XM_005248682.3 XM_005248683.3 XM_005248684.3 XM_011535596.2 XM_011535597.2 XM_011535598.2 XM_011535599.2 XM_011535600.2 XM_017010493.1 XM_017010494.1 XM_017010495.1 XM_017010496.1 XM_017010497.1 XM_017010498.1 XM_017010499.1 XM_017010500.1 XM_017010501.1 XM_017010502.1 XM_017010503.1 XM_017010504.1 XM_017010505.1 XM_017010506.1 XM_017010507.1 XM_017010508.1 XM_017010509.1 XM_017010510.1 XM_017010511.1 XM_017010512.1 XM_017010513.1 XM_017010514.1 XM_017010515.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370704 c.419C>T p.A140V Chr.6: 89612273-89612273 on Build GRCh38
6836883 c.430G>A p.A144T Chr.6: 89612284-89612284 on Build GRCh38
5871219 c.493T>C p.S165P Chr.6: 89612347-89612347 on Build GRCh38
6567549 c.507C>G p.L169L Chr.6: 89612361-89612361 on Build GRCh38
5078650 c.418-10T>C p.? Chr.6: 89612262-89612262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980836156 Chr.6: 89612397-89612397 on Build GRCh38
rs373883234 Chr.6: 89612306-89612307 on Build GRCh38
rs201654033 Chr.6: 89612361-89612361 on Build GRCh38
rs755003542 Chr.6: 89612396-89612396 on Build GRCh38
rs377367109 Chr.6: 89612325-89612325 on Build GRCh38
rs780594879 Chr.6: 89612250-89612250 on Build GRCh38
rs770983807 Chr.6: 89612306-89612306 on Build GRCh38
rs189493829 Chr.6: 89612359-89612359 on Build GRCh38
rs371115312 Chr.6: 89612343-89612343 on Build GRCh38
rs757891765 Chr.6: 89612244-89612244 on Build GRCh38
rs879053748 Chr.6: 89612350-89612350 on Build GRCh38
rs865895910 Chr.6: 89612315-89612315 on Build GRCh38
rs894267475 Chr.6: 89612317-89612317 on Build GRCh38
rs753650260 Chr.6: 89612326-89612326 on Build GRCh38
rs781476025 Chr.6: 89612274-89612274 on Build GRCh38
rs542360344 Chr.6: 89612293-89612293 on Build GRCh38
rs769153391 Chr.6: 89612273-89612273 on Build GRCh38
rs868585931 Chr.6: 89612303-89612306 on Build GRCh38
rs904210793 Chr.6: 89612297-89612297 on Build GRCh38
rs371829055 Chr.6: 89612380-89612380 on Build GRCh38
rs181602896 Chr.6: 89612406-89612406 on Build GRCh38
rs563658021 Chr.6: 89612324-89612324 on Build GRCh38
rs769962774 Chr.6: 89612287-89612287 on Build GRCh38
rs778465327 Chr.6: 89612241-89612241 on Build GRCh38
rs749406437 Chr.6: 89612401-89612401 on Build GRCh38
rs2073272 Chr.6: 89612236-89612236 on Build GRCh38
rs761744665 Chr.6: 89612328-89612328 on Build GRCh38
rs530838661 Chr.6: 89612329-89612329 on Build GRCh38
rs368161206 Chr.6: 89612411-89612411 on Build GRCh38
rs763917096 Chr.6: 89612224-89612224 on Build GRCh38
rs774494829 Chr.6: 89612320-89612320 on Build GRCh38
rs144577628 Chr.6: 89612231-89612231 on Build GRCh38
rs374381779 Chr.6: 89612243-89612243 on Build GRCh38
rs888121359 Chr.6: 89612344-89612344 on Build GRCh38
rs754461983 Chr.6: 89612403-89612406 on Build GRCh38
rs1048085572 Chr.6: 89612337-89612337 on Build GRCh38
rs767267407 Chr.6: 89612283-89612283 on Build GRCh38
rs757918748 Chr.6: 89612348-89612348 on Build GRCh38
rs771200624 Chr.6: 89612404-89612404 on Build GRCh38
rs552443525 Chr.6: 89612353-89612353 on Build GRCh38
rs751081085 Chr.6: 89612352-89612352 on Build GRCh38
rs747533509 Chr.6: 89612251-89612251 on Build GRCh38
rs748534196 Chr.6: 89612277-89612277 on Build GRCh38
rs376144287 Chr.6: 89612351-89612351 on Build GRCh38

View Full Product Details