Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 44372619-44372880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMKMT
Entrez Gene ID
79823
Gene Name
calmodulin-lysine N-methyltransferase
Gene Aliases
C2orf34, CLNMT, CaM KMT, Cam, KMT
UniGene
Hs.468349
Gene Chromosome Location
Chr.2: 44361714-44774388 on Build GRCh38
Transcripts
NM_024766.4 XM_011533111.2 XM_017004970.1 XM_017004971.1 XM_017004972.1 XM_017004973.1 XM_017004974.1 XM_017004975.1 XM_017004976.1 XM_017004977.1 XM_017004978.1 XM_017004979.1 XM_017004980.1 XM_017004981.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094351 c.214T>C p.S72P Chr.2: 44372791-44372791 on Build GRCh38
6488187 c.270A>T p.Q90H Chr.2: 44372847-44372847 on Build GRCh38
4779899 c.227G>T p.G76V Chr.2: 44372804-44372804 on Build GRCh38
3581797 c.289C>G p.P97A Chr.2: 44372866-44372866 on Build GRCh38
5435708 c.215C>G p.S72* Chr.2: 44372792-44372792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570232376 Chr.2: 44372793-44372793 on Build GRCh38
rs779931030 Chr.2: 44372716-44372716 on Build GRCh38
rs138924512 Chr.2: 44372734-44372734 on Build GRCh38
rs577513496 Chr.2: 44372730-44372730 on Build GRCh38
rs542332931 Chr.2: 44372827-44372827 on Build GRCh38
rs765762694 Chr.2: 44372683-44372683 on Build GRCh38
rs772090739 Chr.2: 44372737-44372737 on Build GRCh38
rs61744230 Chr.2: 44372799-44372799 on Build GRCh38
rs755885768 Chr.2: 44372797-44372797 on Build GRCh38
rs766580975 Chr.2: 44372748-44372748 on Build GRCh38
rs774309366 Chr.2: 44372733-44372733 on Build GRCh38
rs1041445322 Chr.2: 44372646-44372646 on Build GRCh38
rs748532181 Chr.2: 44372731-44372731 on Build GRCh38
rs766670848 Chr.2: 44372679-44372679 on Build GRCh38
rs759849827 Chr.2: 44372852-44372852 on Build GRCh38
rs369307588 Chr.2: 44372754-44372754 on Build GRCh38
rs749093011 Chr.2: 44372723-44372723 on Build GRCh38
rs201610490 Chr.2: 44372755-44372755 on Build GRCh38
rs562008225 Chr.2: 44372845-44372845 on Build GRCh38
rs376001186 Chr.2: 44372674-44372674 on Build GRCh38
rs781027302 Chr.2: 44372696-44372696 on Build GRCh38
rs756743744 Chr.2: 44372759-44372759 on Build GRCh38
rs945626575 Chr.2: 44372753-44372753 on Build GRCh38
rs962063109 Chr.2: 44372795-44372795 on Build GRCh38
rs777039644 Chr.2: 44372749-44372749 on Build GRCh38
rs779827140 Chr.2: 44372798-44372798 on Build GRCh38
rs778738809 Chr.2: 44372814-44372814 on Build GRCh38
rs368894831 Chr.2: 44372769-44372769 on Build GRCh38
rs751080201 Chr.2: 44372685-44372685 on Build GRCh38
rs773434568 Chr.2: 44372738-44372738 on Build GRCh38
rs759924384 Chr.2: 44372750-44372750 on Build GRCh38
rs745472830 Chr.2: 44372707-44372707 on Build GRCh38
rs999961520 Chr.2: 44372830-44372830 on Build GRCh38
rs772142079 Chr.2: 44372829-44372829 on Build GRCh38
rs760819119 Chr.2: 44372746-44372746 on Build GRCh38
rs924313283 Chr.2: 44372695-44372695 on Build GRCh38
rs755504979 Chr.2: 44372681-44372681 on Build GRCh38
rs750107198 Chr.2: 44372791-44372791 on Build GRCh38
rs754758348 Chr.2: 44372804-44372804 on Build GRCh38
rs1015004032 Chr.2: 44372768-44372768 on Build GRCh38
rs756760858 Chr.2: 44372686-44372686 on Build GRCh38
rs747080376 Chr.2: 44372844-44372844 on Build GRCh38
rs776881690 Chr.2: 44372847-44372847 on Build GRCh38
rs755989682 Chr.2: 44372712-44372712 on Build GRCh38
rs894358597 Chr.2: 44372736-44372736 on Build GRCh38
rs767669537 Chr.2: 44372671-44372671 on Build GRCh38
rs777893861 Chr.2: 44372831-44372831 on Build GRCh38

View Full Product Details