Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8261899-8262170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS4
Entrez Gene ID
79603
Gene Name
ceramide synthase 4
Gene Aliases
LASS4, Trh1
UniGene
Hs.515111
Gene Chromosome Location
Chr.19: 8209329-8262433 on Build GRCh38
Transcripts
NM_024552.2 XM_011528290.2 XM_011528291.2 XM_011528292.2 XM_011528293.2 XM_011528294.2 XM_011528295.2 XM_017027303.1 XM_017027304.1 XM_017027305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839748 c.1025G>C p.S342T Chr.19: 8261949-8261949 on Build GRCh38
2730173 c.1033G>C p.E345Q Chr.19: 8261957-8261957 on Build GRCh38
6412241 c.1061C>A p.A354E Chr.19: 8261985-8261985 on Build GRCh38
7091065 c.1021C>A p.R341S Chr.19: 8261945-8261945 on Build GRCh38
2730178 c.1141C>T p.R381W Chr.19: 8262065-8262065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758697674 Chr.19: 8261941-8261942 on Build GRCh38
rs377703774 Chr.19: 8262136-8262137 on Build GRCh38
rs764705410 Chr.19: 8261984-8261984 on Build GRCh38
rs745991264 Chr.19: 8262113-8262113 on Build GRCh38
rs746932191 Chr.19: 8261954-8261954 on Build GRCh38
rs377204315 Chr.19: 8261985-8261985 on Build GRCh38
rs757679229 Chr.19: 8262036-8262036 on Build GRCh38
rs778564076 Chr.19: 8261989-8261989 on Build GRCh38
rs17160348 Chr.19: 8261982-8261982 on Build GRCh38
rs778649897 Chr.19: 8262082-8262083 on Build GRCh38
rs768478467 Chr.19: 8262079-8262080 on Build GRCh38
rs768303981 Chr.19: 8262017-8262017 on Build GRCh38
rs576348408 Chr.19: 8262016-8262016 on Build GRCh38
rs17160349 Chr.19: 8262060-8262060 on Build GRCh38
rs554420170 Chr.19: 8261979-8261979 on Build GRCh38
rs534378217 Chr.19: 8261924-8261924 on Build GRCh38
rs199893919 Chr.19: 8262010-8262010 on Build GRCh38
rs918186126 Chr.19: 8262082-8262082 on Build GRCh38
rs375361915 Chr.19: 8262051-8262051 on Build GRCh38
rs1027006474 Chr.19: 8261955-8261955 on Build GRCh38
rs775840159 Chr.19: 8262116-8262116 on Build GRCh38
rs878862385 Chr.19: 8262136-8262136 on Build GRCh38
rs772244323 Chr.19: 8262048-8262048 on Build GRCh38
rs777261835 Chr.19: 8262065-8262065 on Build GRCh38
rs762345692 Chr.19: 8262066-8262066 on Build GRCh38
rs779927057 Chr.19: 8261951-8261951 on Build GRCh38
rs370795271 Chr.19: 8262027-8262027 on Build GRCh38
rs970090617 Chr.19: 8261977-8261977 on Build GRCh38
rs754856297 Chr.19: 8262098-8262098 on Build GRCh38
rs752060566 Chr.19: 8261979-8261980 on Build GRCh38
rs753077018 Chr.19: 8261934-8261936 on Build GRCh38
rs772061570 Chr.19: 8261950-8261950 on Build GRCh38
rs759746399 Chr.19: 8261980-8261980 on Build GRCh38
rs892884825 Chr.19: 8262126-8262126 on Build GRCh38
rs763129211 Chr.19: 8261973-8261973 on Build GRCh38
rs745502911 Chr.19: 8261946-8261946 on Build GRCh38
rs909260186 Chr.19: 8262073-8262073 on Build GRCh38
rs772695825 Chr.19: 8261990-8261990 on Build GRCh38
rs779651444 Chr.19: 8262070-8262077 on Build GRCh38
rs368707575 Chr.19: 8261925-8261925 on Build GRCh38
rs751145645 Chr.19: 8262094-8262094 on Build GRCh38
rs773195742 Chr.19: 8261972-8261972 on Build GRCh38
rs759779063 Chr.19: 8262043-8262043 on Build GRCh38
rs776804362 Chr.19: 8262127-8262127 on Build GRCh38
rs778178926 Chr.19: 8261954-8261954 on Build GRCh38
rs778998101 Chr.19: 8262112-8262112 on Build GRCh38
rs556478251 Chr.19: 8261986-8261986 on Build GRCh38
rs1045702346 Chr.19: 8262109-8262109 on Build GRCh38
rs747961029 Chr.19: 8262096-8262097 on Build GRCh38
rs767852651 Chr.19: 8261921-8261921 on Build GRCh38
rs753352606 Chr.19: 8261935-8261935 on Build GRCh38
rs756694466 Chr.19: 8261938-8261938 on Build GRCh38
rs761154765 Chr.19: 8262059-8262059 on Build GRCh38
rs780015199 Chr.19: 8262014-8262014 on Build GRCh38
rs747191982 Chr.19: 8262119-8262119 on Build GRCh38
rs868276266 Chr.19: 8262013-8262013 on Build GRCh38
rs1045216171 Chr.19: 8261961-8261961 on Build GRCh38
rs36259 Chr.19: 8262020-8262020 on Build GRCh38
rs867844937 Chr.19: 8262078-8262078 on Build GRCh38
rs770972582 Chr.19: 8261974-8261974 on Build GRCh38
rs1044547 Chr.19: 8262115-8262115 on Build GRCh38
rs748017894 Chr.19: 8262022-8262022 on Build GRCh38
rs138254504 Chr.19: 8261983-8261983 on Build GRCh38
rs927801585 Chr.19: 8262129-8262129 on Build GRCh38
rs771355894 Chr.19: 8262074-8262074 on Build GRCh38
rs540801328 Chr.19: 8262077-8262077 on Build GRCh38
rs772861121 Chr.19: 8262137-8262137 on Build GRCh38
rs978902204 Chr.19: 8262055-8262055 on Build GRCh38
rs774345353 Chr.19: 8262033-8262033 on Build GRCh38
rs756849017 Chr.19: 8261988-8261988 on Build GRCh38
rs768589461 Chr.19: 8261965-8261965 on Build GRCh38
rs371254466 Chr.19: 8261941-8261941 on Build GRCh38
rs751288236 Chr.19: 8262144-8262144 on Build GRCh38
rs76848957 Chr.19: 8262143-8262143 on Build GRCh38
rs766174359 Chr.19: 8262091-8262091 on Build GRCh38
rs1010342388 Chr.19: 8262138-8262138 on Build GRCh38
rs549602365 Chr.19: 8262139-8262139 on Build GRCh38
rs765797102 Chr.19: 8262142-8262142 on Build GRCh38
rs375090438 Chr.19: 8261967-8261967 on Build GRCh38
rs748741148 Chr.19: 8262070-8262071 on Build GRCh38
rs758154980 Chr.19: 8262083-8262083 on Build GRCh38
rs748118632 Chr.19: 8262107-8262107 on Build GRCh38
rs756000347 Chr.19: 8262111-8262111 on Build GRCh38
rs563942801 Chr.19: 8262100-8262100 on Build GRCh38

View Full Product Details