Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 126019600-126019857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM12
Entrez Gene ID
8038
Gene Name
ADAM metallopeptidase domain 12
Gene Aliases
ADAM12-OT1, CAR10, MCMP, MCMPMltna, MLTN, MLTNA
UniGene
Hs.594351
Gene Chromosome Location
Chr.10: 126012385-126388558 on Build GRCh38
Transcripts
NM_001288973.1 NM_003474.5 XM_017016705.1 XM_017016706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
352287 c.2630C>A p.P877Q Chr.10: 126019734-126019734 on Build GRCh38
1703233 c.2647G>A p.G883R Chr.10: 126019717-126019717 on Build GRCh38
308862 c.2543C>T p.T848I Chr.10: 126019821-126019821 on Build GRCh38
3666713 c.2577T>A p.P859P Chr.10: 126019787-126019787 on Build GRCh38
3666711 c.2625G>A p.R875R Chr.10: 126019739-126019739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754717910 Chr.10: 126019676-126019676 on Build GRCh38
rs112322315 Chr.10: 126019815-126019815 on Build GRCh38
rs765897289 Chr.10: 126019754-126019754 on Build GRCh38
rs748051393 Chr.10: 126019688-126019688 on Build GRCh38
rs908412831 Chr.10: 126019636-126019636 on Build GRCh38
rs774705109 Chr.10: 126019821-126019821 on Build GRCh38
rs1042829893 Chr.10: 126019646-126019646 on Build GRCh38
rs150268596 Chr.10: 126019702-126019702 on Build GRCh38
rs773811966 Chr.10: 126019738-126019738 on Build GRCh38
rs375345790 Chr.10: 126019772-126019772 on Build GRCh38
rs762300924 Chr.10: 126019735-126019735 on Build GRCh38
rs756219749 Chr.10: 126019817-126019817 on Build GRCh38
rs758251996 Chr.10: 126019787-126019787 on Build GRCh38
rs749549938 Chr.10: 126019717-126019717 on Build GRCh38
rs760158795 Chr.10: 126019746-126019746 on Build GRCh38
rs141115475 Chr.10: 126019699-126019699 on Build GRCh38
rs141746959 Chr.10: 126019731-126019731 on Build GRCh38
rs764997787 Chr.10: 126019779-126019779 on Build GRCh38
rs544198566 Chr.10: 126019620-126019620 on Build GRCh38
rs778603998 Chr.10: 126019678-126019678 on Build GRCh38
rs56409749 Chr.10: 126019808-126019808 on Build GRCh38
rs200515505 Chr.10: 126019689-126019689 on Build GRCh38
rs1005756061 Chr.10: 126019718-126019718 on Build GRCh38
rs767909233 Chr.10: 126019737-126019737 on Build GRCh38
rs138915748 Chr.10: 126019711-126019711 on Build GRCh38
rs777134947 Chr.10: 126019635-126019635 on Build GRCh38
rs903389293 Chr.10: 126019831-126019831 on Build GRCh38
rs752707322 Chr.10: 126019720-126019720 on Build GRCh38
rs749458440 Chr.10: 126019820-126019820 on Build GRCh38
rs753535686 Chr.10: 126019663-126019663 on Build GRCh38
rs374420879 Chr.10: 126019818-126019818 on Build GRCh38
rs376942222 Chr.10: 126019785-126019785 on Build GRCh38
rs748432789 Chr.10: 126019835-126019835 on Build GRCh38
rs770041154 Chr.10: 126019710-126019710 on Build GRCh38
rs765313755 Chr.10: 126019658-126019661 on Build GRCh38
rs758654497 Chr.10: 126019802-126019803 on Build GRCh38
rs769071080 Chr.10: 126019719-126019719 on Build GRCh38
rs199632862 Chr.10: 126019788-126019788 on Build GRCh38
rs370639488 Chr.10: 126019661-126019661 on Build GRCh38
rs753449687 Chr.10: 126019761-126019761 on Build GRCh38
rs563177267 Chr.10: 126019626-126019626 on Build GRCh38
rs941274896 Chr.10: 126019654-126019654 on Build GRCh38

View Full Product Details