Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 904019-904292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAP1
Entrez Gene ID
11033
Gene Name
ArfGAP with dual PH domains 1
Gene Aliases
CENTA1, GCS1L, p42IP4
UniGene
Hs.602573
Gene Chromosome Location
Chr.7: 897900-955407 on Build GRCh38
Transcripts
NM_001284308.1 NM_001284309.1 NM_001284310.1 NM_001284311.1 NM_006869.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122897 c.616C>A p.R206S Chr.7: 904158-904158 on Build GRCh38
1165429 c.616C>T p.R206C Chr.7: 904158-904158 on Build GRCh38
4871636 c.669T>C p.H223H Chr.7: 904138-904138 on Build GRCh38
6479838 c.561G>C p.K187N Chr.7: 904246-904246 on Build GRCh38
6012600 c.639G>A p.E213E Chr.7: 904135-904135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757675790 Chr.7: 904080-904080 on Build GRCh38
rs62430821 Chr.7: 904062-904062 on Build GRCh38
rs76805149 Chr.7: 904099-904099 on Build GRCh38
rs74366959 Chr.7: 904255-904255 on Build GRCh38
rs116145502 Chr.7: 904075-904075 on Build GRCh38
rs76984741 Chr.7: 904257-904257 on Build GRCh38
rs771992996 Chr.7: 904091-904091 on Build GRCh38
rs1040719921 Chr.7: 904057-904057 on Build GRCh38
rs775317996 Chr.7: 904171-904171 on Build GRCh38
rs551847885 Chr.7: 904055-904055 on Build GRCh38
rs774656853 Chr.7: 904181-904181 on Build GRCh38
rs746997218 Chr.7: 904141-904141 on Build GRCh38
rs564297398 Chr.7: 904046-904046 on Build GRCh38
rs548144013 Chr.7: 904068-904068 on Build GRCh38
rs200044511 Chr.7: 904132-904132 on Build GRCh38
rs945204362 Chr.7: 904253-904253 on Build GRCh38
rs560336903 Chr.7: 904123-904123 on Build GRCh38
rs373997978 Chr.7: 904105-904105 on Build GRCh38
rs140582393 Chr.7: 904204-904204 on Build GRCh38
rs369166492 Chr.7: 904086-904086 on Build GRCh38
rs777524048 Chr.7: 904139-904139 on Build GRCh38
rs981816395 Chr.7: 904225-904225 on Build GRCh38
rs769756910 Chr.7: 904218-904218 on Build GRCh38
rs183170940 Chr.7: 904040-904040 on Build GRCh38
rs368329171 Chr.7: 904186-904186 on Build GRCh38
rs780321785 Chr.7: 904131-904131 on Build GRCh38
rs927679820 Chr.7: 904039-904039 on Build GRCh38
rs756511581 Chr.7: 904197-904197 on Build GRCh38
rs371597954 Chr.7: 904113-904113 on Build GRCh38
rs201134549 Chr.7: 904270-904270 on Build GRCh38
rs74656328 Chr.7: 904048-904048 on Build GRCh38
rs142389101 Chr.7: 904138-904138 on Build GRCh38
rs146488513 Chr.7: 904093-904093 on Build GRCh38
rs751863806 Chr.7: 904079-904079 on Build GRCh38
rs764781696 Chr.7: 904104-904104 on Build GRCh38
rs138852813 Chr.7: 904245-904245 on Build GRCh38
rs138262967 Chr.7: 904215-904215 on Build GRCh38
rs553173230 Chr.7: 904228-904228 on Build GRCh38
rs371063602 Chr.7: 904100-904100 on Build GRCh38
rs370835412 Chr.7: 904158-904158 on Build GRCh38
rs897028813 Chr.7: 904159-904159 on Build GRCh38
rs781625991 Chr.7: 904118-904118 on Build GRCh38
rs749155711 Chr.7: 904268-904268 on Build GRCh38
rs369464000 Chr.7: 904061-904061 on Build GRCh38
rs755011413 Chr.7: 904200-904200 on Build GRCh38
rs769811232 Chr.7: 904242-904242 on Build GRCh38
rs776497998 Chr.7: 904153-904153 on Build GRCh38
rs567979929 Chr.7: 904069-904069 on Build GRCh38
rs899407573 Chr.7: 904056-904086 on Build GRCh38
rs764472130 Chr.7: 904112-904112 on Build GRCh38
rs780011043 Chr.7: 904264-904264 on Build GRCh38
rs1006906229 Chr.7: 904201-904201 on Build GRCh38
rs773757384 Chr.7: 904236-904236 on Build GRCh38
rs775231609 Chr.7: 904220-904220 on Build GRCh38
rs762399070 Chr.7: 904185-904185 on Build GRCh38
rs770564311 Chr.7: 904146-904146 on Build GRCh38
rs573403038 Chr.7: 904092-904092 on Build GRCh38
rs377425159 Chr.7: 904157-904157 on Build GRCh38
rs191140366 Chr.7: 904195-904195 on Build GRCh38
rs570658393 Chr.7: 904077-904077 on Build GRCh38
rs149515070 Chr.7: 904254-904254 on Build GRCh38
rs746136322 Chr.7: 904082-904082 on Build GRCh38
rs139899830 Chr.7: 904106-904106 on Build GRCh38
rs781245053 Chr.7: 904081-904081 on Build GRCh38
rs376055866 Chr.7: 904216-904216 on Build GRCh38
rs202209725 Chr.7: 904261-904261 on Build GRCh38
rs1037895644 Chr.7: 904049-904079 on Build GRCh38
rs370343329 Chr.7: 904114-904114 on Build GRCh38
rs748976827 Chr.7: 904226-904226 on Build GRCh38
rs10282526 Chr.7: 904074-904074 on Build GRCh38
rs61729638 Chr.7: 904177-904177 on Build GRCh38
rs539684769 Chr.7: 904084-904084 on Build GRCh38
rs145071792 Chr.7: 904243-904243 on Build GRCh38
rs971594412 Chr.7: 904217-904217 on Build GRCh38
rs150403059 Chr.7: 904207-904207 on Build GRCh38
rs909811133 Chr.7: 904147-904147 on Build GRCh38
rs767970805 Chr.7: 904194-904194 on Build GRCh38
rs766566324 Chr.7: 904198-904198 on Build GRCh38
rs567634941 Chr.7: 904062-904092 on Build GRCh38
rs373464798 Chr.7: 904127-904127 on Build GRCh38
rs374403101 Chr.7: 904180-904180 on Build GRCh38
rs61730955 Chr.7: 904231-904231 on Build GRCh38
rs34320061 Chr.7: 904176-904176 on Build GRCh38
rs370538517 Chr.7: 904206-904206 on Build GRCh38

View Full Product Details