Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22638777-22639008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QA
Entrez Gene ID
712
Gene Name
complement C1q A chain
Gene Aliases
-
UniGene
Hs.632379
Gene Chromosome Location
Chr.1: 22636625-22639682 on Build GRCh38
Transcripts
NM_015991.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3484921 c.334C>T p.Q112* Chr.1: 22639003-22639003 on Build GRCh38
1687170 c.232A>C p.N78H Chr.1: 22638901-22638901 on Build GRCh38
3484919 c.214G>A p.E72K Chr.1: 22638883-22638883 on Build GRCh38
5659016 c.220G>A p.G74R Chr.1: 22638889-22638889 on Build GRCh38
3484918 c.202G>A p.G68R Chr.1: 22638871-22638871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781752563 Chr.1: 22638923-22638923 on Build GRCh38
rs180679721 Chr.1: 22638964-22638964 on Build GRCh38
rs759712266 Chr.1: 22638978-22638978 on Build GRCh38
rs528301944 Chr.1: 22638881-22638881 on Build GRCh38
rs902565316 Chr.1: 22638943-22638943 on Build GRCh38
rs1023182910 Chr.1: 22638928-22638928 on Build GRCh38
rs772544683 Chr.1: 22638848-22638848 on Build GRCh38
rs904647605 Chr.1: 22638813-22638813 on Build GRCh38
rs766694409 Chr.1: 22638975-22638975 on Build GRCh38
rs372914310 Chr.1: 22638933-22638933 on Build GRCh38
rs998837784 Chr.1: 22638830-22638830 on Build GRCh38
rs960202389 Chr.1: 22638854-22638854 on Build GRCh38
rs761744728 Chr.1: 22638864-22638864 on Build GRCh38
rs748582147 Chr.1: 22638847-22638847 on Build GRCh38
rs755011795 Chr.1: 22638922-22638922 on Build GRCh38
rs769802299 Chr.1: 22638950-22638950 on Build GRCh38
rs766456234 Chr.1: 22638888-22638888 on Build GRCh38
rs762772392 Chr.1: 22638972-22638972 on Build GRCh38
rs748632438 Chr.1: 22638927-22638927 on Build GRCh38
rs867255630 Chr.1: 22638946-22638946 on Build GRCh38
rs749595647 Chr.1: 22638934-22638934 on Build GRCh38
rs932927116 Chr.1: 22638812-22638812 on Build GRCh38
rs779568085 Chr.1: 22638939-22638939 on Build GRCh38
rs775077199 Chr.1: 22638835-22638835 on Build GRCh38
rs774041017 Chr.1: 22638811-22638811 on Build GRCh38
rs771758729 Chr.1: 22638935-22638935 on Build GRCh38
rs172378 Chr.1: 22638945-22638945 on Build GRCh38
rs765090332 Chr.1: 22638865-22638865 on Build GRCh38
rs751668628 Chr.1: 22638894-22638894 on Build GRCh38
rs567106778 Chr.1: 22638940-22638940 on Build GRCh38
rs377231186 Chr.1: 22638942-22638942 on Build GRCh38
rs920195980 Chr.1: 22638802-22638802 on Build GRCh38
rs992778617 Chr.1: 22638874-22638874 on Build GRCh38
rs751720642 Chr.1: 22638976-22638976 on Build GRCh38
rs776416316 Chr.1: 22638855-22638855 on Build GRCh38
rs758718727 Chr.1: 22638886-22638886 on Build GRCh38
rs745489495 Chr.1: 22638822-22638822 on Build GRCh38
rs112251360 Chr.1: 22638831-22638831 on Build GRCh38

View Full Product Details