Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3902374-3902645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATCAY
Entrez Gene ID
85300
Gene Name
ATCAY, caytaxin
Gene Aliases
BNIP-H, CLAC
UniGene
Hs.418055
Gene Chromosome Location
Chr.19: 3880620-3928082 on Build GRCh38
Transcripts
NM_033064.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6407929 c.130A>G p.T44A Chr.19: 3902539-3902539 on Build GRCh38
4639043 c.109C>T p.L37F Chr.19: 3902518-3902518 on Build GRCh38
6407928 c.130A>G p.T44A Chr.19: 3902539-3902539 on Build GRCh38
355855 c.136T>A p.S46T Chr.19: 3902545-3902545 on Build GRCh38
5416488 c.119C>T p.P40L Chr.19: 3902528-3902528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746877635 Chr.19: 3902561-3902561 on Build GRCh38
rs147031440 Chr.19: 3902507-3902507 on Build GRCh38
rs1019745727 Chr.19: 3902428-3902428 on Build GRCh38
rs754044132 Chr.19: 3902557-3902557 on Build GRCh38
rs747675432 Chr.19: 3902581-3902581 on Build GRCh38
rs549656846 Chr.19: 3902528-3902528 on Build GRCh38
rs780011050 Chr.19: 3902559-3902559 on Build GRCh38
rs191872542 Chr.19: 3902396-3902396 on Build GRCh38
rs777920713 Chr.19: 3902453-3902453 on Build GRCh38
rs984000548 Chr.19: 3902505-3902505 on Build GRCh38
rs200614118 Chr.19: 3902505-3902505 on Build GRCh38
rs748959413 Chr.19: 3902595-3902595 on Build GRCh38
rs766539565 Chr.19: 3902584-3902584 on Build GRCh38
rs764352635 Chr.19: 3902534-3902534 on Build GRCh38
rs1009085062 Chr.19: 3902399-3902399 on Build GRCh38
rs369421443 Chr.19: 3902483-3902483 on Build GRCh38
rs564270693 Chr.19: 3902437-3902437 on Build GRCh38
rs377313970 Chr.19: 3902529-3902529 on Build GRCh38
rs207476986 Chr.19: 3902508-3902508 on Build GRCh38
rs763273815 Chr.19: 3902506-3902506 on Build GRCh38
rs745659774 Chr.19: 3902455-3902455 on Build GRCh38
rs769474716 Chr.19: 3902504-3902504 on Build GRCh38
rs982222017 Chr.19: 3902572-3902572 on Build GRCh38
rs540999842 Chr.19: 3902431-3902431 on Build GRCh38
rs754962968 Chr.19: 3902563-3902563 on Build GRCh38
rs1028100895 Chr.19: 3902462-3902462 on Build GRCh38
rs559759678 Chr.19: 3902558-3902558 on Build GRCh38
rs529417443 Chr.19: 3902526-3902526 on Build GRCh38
rs1054010029 Chr.19: 3902618-3902618 on Build GRCh38
rs760048671 Chr.19: 3902520-3902520 on Build GRCh38
rs758314365 Chr.19: 3902456-3902456 on Build GRCh38
rs774788713 Chr.19: 3902509-3902509 on Build GRCh38
rs747684170 Chr.19: 3902502-3902502 on Build GRCh38
rs577702656 Chr.19: 3902452-3902452 on Build GRCh38
rs374321481 Chr.19: 3902593-3902593 on Build GRCh38
rs369888276 Chr.19: 3902571-3902571 on Build GRCh38
rs752804093 Chr.19: 3902439-3902439 on Build GRCh38
rs768227102 Chr.19: 3902489-3902489 on Build GRCh38
rs1009245200 Chr.19: 3902407-3902407 on Build GRCh38
rs369271502 Chr.19: 3902458-3902458 on Build GRCh38
rs776213982 Chr.19: 3902494-3902494 on Build GRCh38

View Full Product Details