Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8207017-8207276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKB
Entrez Gene ID
9212
Gene Name
aurora kinase B
Gene Aliases
AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2
UniGene
Hs.442658
Gene Chromosome Location
Chr.17: 8204731-8210768 on Build GRCh38
Transcripts
NM_001256834.2 NM_001284526.1 NM_001313950.1 NM_001313951.1 NM_001313952.1 NM_001313953.1 NM_001313954.1 NM_001313955.1 NM_004217.3 NR_132730.1 NR_132731.1 XM_011524070.2 XM_011524072.2 XM_017025307.1 XM_017025308.1 XM_017025309.1 XM_017025310.1 XM_017025311.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4172216 c.334C>T p.Q112* Chr.17: 8207240-8207240 on Build GRCh38
6002460 c.351C>T p.G117G Chr.17: 8207223-8207223 on Build GRCh38
5427486 c.348G>A p.E116E Chr.17: 8207226-8207226 on Build GRCh38
3523935 c.303C>T p.F101F Chr.17: 8207271-8207271 on Build GRCh38
986227 c.378C>T p.I126I Chr.17: 8207196-8207196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053838144 Chr.17: 8207127-8207127 on Build GRCh38
rs949493728 Chr.17: 8207094-8207094 on Build GRCh38
rs369061154 Chr.17: 8207099-8207099 on Build GRCh38
rs746022827 Chr.17: 8207181-8207181 on Build GRCh38
rs768744426 Chr.17: 8207205-8207205 on Build GRCh38
rs762386906 Chr.17: 8207223-8207223 on Build GRCh38
rs765662740 Chr.17: 8207128-8207128 on Build GRCh38
rs766172470 Chr.17: 8207242-8207242 on Build GRCh38
rs777632679 Chr.17: 8207173-8207173 on Build GRCh38
rs936755074 Chr.17: 8207106-8207106 on Build GRCh38
rs750077492 Chr.17: 8207229-8207231 on Build GRCh38
rs751088873 Chr.17: 8207256-8207256 on Build GRCh38
rs148364684 Chr.17: 8207232-8207232 on Build GRCh38
rs747133622 Chr.17: 8207199-8207199 on Build GRCh38
rs972123672 Chr.17: 8207051-8207051 on Build GRCh38
rs573362429 Chr.17: 8207095-8207095 on Build GRCh38
rs765789739 Chr.17: 8207138-8207138 on Build GRCh38
rs937138967 Chr.17: 8207252-8207252 on Build GRCh38
rs777080470 Chr.17: 8207219-8207219 on Build GRCh38
rs766111724 Chr.17: 8207152-8207152 on Build GRCh38
rs751176876 Chr.17: 8207145-8207145 on Build GRCh38
rs749976036 Chr.17: 8207131-8207131 on Build GRCh38
rs113006369 Chr.17: 8207175-8207175 on Build GRCh38
rs909428128 Chr.17: 8207047-8207047 on Build GRCh38
rs970657376 Chr.17: 8207053-8207053 on Build GRCh38
rs754932503 Chr.17: 8207151-8207151 on Build GRCh38
rs773681102 Chr.17: 8207226-8207226 on Build GRCh38
rs942272401 Chr.17: 8207048-8207048 on Build GRCh38
rs765462146 Chr.17: 8207225-8207225 on Build GRCh38
rs192781364 Chr.17: 8207162-8207162 on Build GRCh38
rs980792521 Chr.17: 8207066-8207066 on Build GRCh38
rs748122245 Chr.17: 8207153-8207153 on Build GRCh38
rs34360875 Chr.17: 8207147-8207147 on Build GRCh38
rs919237127 Chr.17: 8207072-8207072 on Build GRCh38
rs141915374 Chr.17: 8207196-8207196 on Build GRCh38

View Full Product Details