Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3664007-3664278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART1
Entrez Gene ID
417
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
ART2, ARTC1, CD296, RT6
UniGene
Hs.382188
Gene Chromosome Location
Chr.11: 3645110-3665595 on Build GRCh38
Transcripts
NM_004314.2 XM_011520114.2 XM_017017763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3447113 c.973G>A p.G325S Chr.11: 3664178-3664178 on Build GRCh38
7311433 c.891G>T p.M297I Chr.11: 3664096-3664096 on Build GRCh38
6605340 c.956C>T p.A319V Chr.11: 3664161-3664161 on Build GRCh38
6605334 c.922_923insTGC p.L312_W313insL Chr.11: 3664127-3664128 on Build GRCh38
4622065 c.918G>T p.W306C Chr.11: 3664123-3664123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140449390 Chr.11: 3664105-3664105 on Build GRCh38
rs36041658 Chr.11: 3664150-3664150 on Build GRCh38
rs199801440 Chr.11: 3664166-3664166 on Build GRCh38
rs777626849 Chr.11: 3664201-3664201 on Build GRCh38
rs545567510 Chr.11: 3664151-3664151 on Build GRCh38
rs141913583 Chr.11: 3664193-3664193 on Build GRCh38
rs372804191 Chr.11: 3664144-3664144 on Build GRCh38
rs1011307452 Chr.11: 3664246-3664246 on Build GRCh38
rs1042417236 Chr.11: 3664197-3664197 on Build GRCh38
rs770524296 Chr.11: 3664209-3664209 on Build GRCh38
rs781104873 Chr.11: 3664207-3664208 on Build GRCh38
rs767182207 Chr.11: 3664049-3664049 on Build GRCh38
rs758415710 Chr.11: 3664058-3664058 on Build GRCh38
rs753016617 Chr.11: 3664067-3664067 on Build GRCh38
rs745883802 Chr.11: 3664088-3664088 on Build GRCh38
rs1045684647 Chr.11: 3664199-3664199 on Build GRCh38
rs147582309 Chr.11: 3664095-3664095 on Build GRCh38
rs767672408 Chr.11: 3664163-3664163 on Build GRCh38
rs753860368 Chr.11: 3664186-3664186 on Build GRCh38
rs375438804 Chr.11: 3664089-3664089 on Build GRCh38
rs971857100 Chr.11: 3664140-3664140 on Build GRCh38
rs867003027 Chr.11: 3664218-3664218 on Build GRCh38
rs757424017 Chr.11: 3664074-3664074 on Build GRCh38
rs777730626 Chr.11: 3664060-3664060 on Build GRCh38
rs760149770 Chr.11: 3664065-3664065 on Build GRCh38
rs779962910 Chr.11: 3664180-3664180 on Build GRCh38
rs756249752 Chr.11: 3664178-3664178 on Build GRCh38
rs771808549 Chr.11: 3664110-3664110 on Build GRCh38
rs1031442714 Chr.11: 3664229-3664229 on Build GRCh38
rs749174565 Chr.11: 3664094-3664094 on Build GRCh38
rs572751063 Chr.11: 3664190-3664190 on Build GRCh38
rs115159160 Chr.11: 3664071-3664071 on Build GRCh38
rs375470433 Chr.11: 3664051-3664051 on Build GRCh38
rs369600232 Chr.11: 3664118-3664118 on Build GRCh38
rs771863585 Chr.11: 3664191-3664191 on Build GRCh38
rs754820651 Chr.11: 3664188-3664188 on Build GRCh38
rs2271582 Chr.11: 3664213-3664213 on Build GRCh38
rs779068215 Chr.11: 3664050-3664051 on Build GRCh38
rs900327784 Chr.11: 3664226-3664226 on Build GRCh38
rs765772376 Chr.11: 3664066-3664066 on Build GRCh38
rs373087910 Chr.11: 3664233-3664233 on Build GRCh38
rs773993156 Chr.11: 3664097-3664097 on Build GRCh38
rs895173248 Chr.11: 3664176-3664176 on Build GRCh38
rs752804745 Chr.11: 3664050-3664053 on Build GRCh38
rs547934498 Chr.11: 3664189-3664189 on Build GRCh38
rs944420905 Chr.11: 3664210-3664210 on Build GRCh38
rs781284713 Chr.11: 3664079-3664079 on Build GRCh38
rs910189822 Chr.11: 3664053-3664053 on Build GRCh38
rs762029532 Chr.11: 3664153-3664153 on Build GRCh38
rs964331658 Chr.11: 3664057-3664057 on Build GRCh38
rs774115524 Chr.11: 3664041-3664041 on Build GRCh38
rs372338750 Chr.11: 3664044-3664044 on Build GRCh38
rs746967675 Chr.11: 3664060-3664061 on Build GRCh38
rs771043956 Chr.11: 3664109-3664110 on Build GRCh38
rs765793639 Chr.11: 3664123-3664123 on Build GRCh38
rs746630142 Chr.11: 3664206-3664206 on Build GRCh38
rs566327340 Chr.11: 3664245-3664245 on Build GRCh38
rs148115951 Chr.11: 3664069-3664069 on Build GRCh38

View Full Product Details