Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149312037-149312310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1L1
Entrez Gene ID
134265
Gene Name
actin filament associated protein 1 like 1
Gene Aliases
-
UniGene
Hs.483793
Gene Chromosome Location
Chr.5: 149271838-149343637 on Build GRCh38
Transcripts
NM_001146337.2 NM_001323062.1 NM_001323063.1 NM_152406.3 XM_011537558.1 XM_017009036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1283299 c.988C>T p.P330S Chr.5: 149312172-149312172 on Build GRCh38
6170042 c.994C>T p.L332F Chr.5: 149312178-149312178 on Build GRCh38
400377 c.1020+1G>A p.? Chr.5: 149312205-149312205 on Build GRCh38
7200560 c.928G>A p.V310I Chr.5: 149312112-149312112 on Build GRCh38
6595741 c.979C>T p.P327S Chr.5: 149312163-149312163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748703166 Chr.5: 149312066-149312066 on Build GRCh38
rs375190446 Chr.5: 149312257-149312257 on Build GRCh38
rs745346002 Chr.5: 149312204-149312204 on Build GRCh38
rs749735233 Chr.5: 149312184-149312184 on Build GRCh38
rs375542540 Chr.5: 149312225-149312225 on Build GRCh38
rs757240110 Chr.5: 149312178-149312178 on Build GRCh38
rs779141585 Chr.5: 149312194-149312194 on Build GRCh38
rs369801283 Chr.5: 149312199-149312199 on Build GRCh38
rs754264450 Chr.5: 149312289-149312289 on Build GRCh38
rs775982070 Chr.5: 149312173-149312173 on Build GRCh38
rs369732949 Chr.5: 149312094-149312094 on Build GRCh38
rs377592650 Chr.5: 149312207-149312207 on Build GRCh38
rs746290546 Chr.5: 149312069-149312069 on Build GRCh38
rs985353506 Chr.5: 149312161-149312161 on Build GRCh38
rs976563930 Chr.5: 149312255-149312255 on Build GRCh38
rs895073226 Chr.5: 149312284-149312284 on Build GRCh38
rs766151234 Chr.5: 149312139-149312139 on Build GRCh38
rs370346755 Chr.5: 149312148-149312148 on Build GRCh38
rs936557985 Chr.5: 149312245-149312245 on Build GRCh38
rs143995508 Chr.5: 149312217-149312217 on Build GRCh38
rs1025112562 Chr.5: 149312144-149312144 on Build GRCh38
rs764680696 Chr.5: 149312268-149312268 on Build GRCh38
rs575334807 Chr.5: 149312137-149312137 on Build GRCh38
rs373449769 Chr.5: 149312096-149312096 on Build GRCh38
rs765074738 Chr.5: 149312121-149312121 on Build GRCh38
rs756372130 Chr.5: 149312265-149312265 on Build GRCh38
rs762556444 Chr.5: 149312230-149312230 on Build GRCh38
rs766026360 Chr.5: 149312237-149312237 on Build GRCh38
rs754645271 Chr.5: 149312157-149312157 on Build GRCh38
rs749395785 Chr.5: 149312175-149312175 on Build GRCh38
rs772262593 Chr.5: 149312206-149312206 on Build GRCh38
rs773974668 Chr.5: 149312240-149312240 on Build GRCh38
rs774494254 Chr.5: 149312085-149312085 on Build GRCh38
rs142577612 Chr.5: 149312084-149312084 on Build GRCh38
rs769776111 Chr.5: 149312224-149312224 on Build GRCh38
rs376857041 Chr.5: 149312118-149312118 on Build GRCh38
rs373211982 Chr.5: 149312067-149312067 on Build GRCh38
rs903823520 Chr.5: 149312062-149312062 on Build GRCh38
rs767272162 Chr.5: 149312260-149312260 on Build GRCh38
rs965091762 Chr.5: 149312250-149312250 on Build GRCh38
rs777964772 Chr.5: 149312073-149312073 on Build GRCh38
rs544299473 Chr.5: 149312119-149312119 on Build GRCh38
rs563099593 Chr.5: 149312136-149312136 on Build GRCh38
rs200632989 Chr.5: 149312226-149312226 on Build GRCh38
rs202121505 Chr.5: 149312163-149312163 on Build GRCh38

View Full Product Details