Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101894738-101895011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL5
Entrez Gene ID
253935
Gene Name
angiopoietin like 5
Gene Aliases
-
UniGene
Hs.318370
Gene Chromosome Location
Chr.11: 101890674-101916522 on Build GRCh38
Transcripts
NM_178127.4 XM_011542735.2 XM_017017466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244939 c.813T>A p.F271L Chr.11: 101894913-101894913 on Build GRCh38
7190492 c.756C>A p.D252E Chr.11: 101894970-101894970 on Build GRCh38
3442614 c.839G>A p.G280E Chr.11: 101894887-101894887 on Build GRCh38
1289441 c.808T>C p.F270L Chr.11: 101894918-101894918 on Build GRCh38
5480806 c.775T>C p.Y259H Chr.11: 101894951-101894951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749530810 Chr.11: 101894905-101894905 on Build GRCh38
rs750584199 Chr.11: 101894939-101894939 on Build GRCh38
rs149099147 Chr.11: 101894897-101894897 on Build GRCh38
rs750117851 Chr.11: 101894856-101894856 on Build GRCh38
rs115960350 Chr.11: 101894774-101894774 on Build GRCh38
rs997387874 Chr.11: 101894813-101894813 on Build GRCh38
rs774846058 Chr.11: 101894917-101894917 on Build GRCh38
rs761839323 Chr.11: 101894865-101894865 on Build GRCh38
rs766085990 Chr.11: 101894952-101894952 on Build GRCh38
rs199727502 Chr.11: 101894852-101894852 on Build GRCh38
rs776540771 Chr.11: 101894864-101894865 on Build GRCh38
rs143196444 Chr.11: 101894921-101894921 on Build GRCh38
rs940564532 Chr.11: 101894948-101894948 on Build GRCh38
rs753384376 Chr.11: 101894842-101894842 on Build GRCh38
rs146549224 Chr.11: 101894878-101894878 on Build GRCh38
rs533067957 Chr.11: 101894812-101894812 on Build GRCh38
rs1019706927 Chr.11: 101894793-101894793 on Build GRCh38
rs761429846 Chr.11: 101894848-101894848 on Build GRCh38
rs575339790 Chr.11: 101894808-101894808 on Build GRCh38
rs1033008707 Chr.11: 101894779-101894779 on Build GRCh38
rs374423446 Chr.11: 101894882-101894882 on Build GRCh38
rs771358332 Chr.11: 101894828-101894828 on Build GRCh38
rs749982132 Chr.11: 101894945-101894945 on Build GRCh38
rs771185921 Chr.11: 101894916-101894916 on Build GRCh38
rs999235551 Chr.11: 101894785-101894785 on Build GRCh38
rs756197428 Chr.11: 101894973-101894973 on Build GRCh38
rs765321536 Chr.11: 101894913-101894913 on Build GRCh38
rs779915483 Chr.11: 101894874-101894874 on Build GRCh38
rs150362257 Chr.11: 101894968-101894968 on Build GRCh38
rs373036063 Chr.11: 101894832-101894832 on Build GRCh38
rs11225051 Chr.11: 101894873-101894873 on Build GRCh38
rs763588629 Chr.11: 101894838-101894838 on Build GRCh38
rs775995082 Chr.11: 101894922-101894922 on Build GRCh38
rs764729251 Chr.11: 101894931-101894931 on Build GRCh38
rs752713598 Chr.11: 101894970-101894970 on Build GRCh38
rs140281758 Chr.11: 101894967-101894967 on Build GRCh38
rs751414863 Chr.11: 101894959-101894959 on Build GRCh38
rs77315074 Chr.11: 101894923-101894923 on Build GRCh38
rs760147168 Chr.11: 101894837-101894837 on Build GRCh38
rs961748906 Chr.11: 101894906-101894906 on Build GRCh38
rs755034899 Chr.11: 101894881-101894881 on Build GRCh38
rs921353165 Chr.11: 101894896-101894896 on Build GRCh38
rs777818263 Chr.11: 101894901-101894901 on Build GRCh38
rs762728136 Chr.11: 101894950-101894950 on Build GRCh38
rs760005579 Chr.11: 101894919-101894919 on Build GRCh38

View Full Product Details