Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10440281-10440554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP2
Entrez Gene ID
80063
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
MCAF2
UniGene
Hs.513343
Gene Chromosome Location
Chr.16: 10386055-10483640 on Build GRCh38
Transcripts
NM_001256160.1 NM_024997.3 NR_045815.1 NR_045816.1 XM_006720953.3 XM_011522666.2 XM_017023705.1 XM_017023706.1 XM_017023707.1 XM_017023708.1 XM_017023710.1 XM_017023711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3887859 c.1114C>T p.Q372* Chr.16: 10440382-10440382 on Build GRCh38
3402052 c.1121G>A p.R374H Chr.16: 10440389-10440389 on Build GRCh38
1718107 c.1096-1G>A p.? Chr.16: 10440363-10440363 on Build GRCh38
6247609 c.1096-2A>T p.? Chr.16: 10440362-10440362 on Build GRCh38
5850131 c.1120C>T p.R374C Chr.16: 10440388-10440388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909062135 Chr.16: 10440392-10440392 on Build GRCh38
rs201209176 Chr.16: 10440504-10440504 on Build GRCh38
rs778176604 Chr.16: 10440452-10440452 on Build GRCh38
rs746602868 Chr.16: 10440391-10440391 on Build GRCh38
rs938394840 Chr.16: 10440494-10440494 on Build GRCh38
rs757147341 Chr.16: 10440319-10440319 on Build GRCh38
rs746625748 Chr.16: 10440336-10440336 on Build GRCh38
rs952803435 Chr.16: 10440375-10440375 on Build GRCh38
rs749835754 Chr.16: 10440365-10440365 on Build GRCh38
rs373506225 Chr.16: 10440337-10440337 on Build GRCh38
rs775822786 Chr.16: 10440496-10440496 on Build GRCh38
rs111917498 Chr.16: 10440522-10440522 on Build GRCh38
rs377544961 Chr.16: 10440406-10440406 on Build GRCh38
rs765216959 Chr.16: 10440325-10440325 on Build GRCh38
rs780715236 Chr.16: 10440349-10440349 on Build GRCh38
rs767736915 Chr.16: 10440388-10440388 on Build GRCh38
rs188503332 Chr.16: 10440419-10440419 on Build GRCh38
rs762657519 Chr.16: 10440510-10440510 on Build GRCh38
rs748474929 Chr.16: 10440350-10440350 on Build GRCh38
rs979900618 Chr.16: 10440453-10440453 on Build GRCh38
rs972027137 Chr.16: 10440417-10440417 on Build GRCh38
rs376683724 Chr.16: 10440475-10440475 on Build GRCh38
rs750434155 Chr.16: 10440330-10440330 on Build GRCh38
rs1024205864 Chr.16: 10440415-10440415 on Build GRCh38
rs573460456 Chr.16: 10440437-10440437 on Build GRCh38
rs1000202281 Chr.16: 10440342-10440342 on Build GRCh38
rs770171425 Chr.16: 10440356-10440356 on Build GRCh38
rs757946523 Chr.16: 10440334-10440334 on Build GRCh38
rs779317955 Chr.16: 10440474-10440474 on Build GRCh38
rs753600904 Chr.16: 10440317-10440317 on Build GRCh38
rs759871311 Chr.16: 10440314-10440314 on Build GRCh38
rs373696008 Chr.16: 10440508-10440508 on Build GRCh38
rs756440051 Chr.16: 10440444-10440444 on Build GRCh38
rs530522396 Chr.16: 10440360-10440360 on Build GRCh38
rs772093256 Chr.16: 10440488-10440488 on Build GRCh38
rs141687995 Chr.16: 10440389-10440389 on Build GRCh38
rs779678020 Chr.16: 10440335-10440335 on Build GRCh38
rs183442276 Chr.16: 10440390-10440390 on Build GRCh38
rs759549507 Chr.16: 10440386-10440386 on Build GRCh38
rs774549900 Chr.16: 10440374-10440374 on Build GRCh38
rs763784264 Chr.16: 10440316-10440316 on Build GRCh38
rs201982427 Chr.16: 10440348-10440348 on Build GRCh38
rs368751131 Chr.16: 10440400-10440400 on Build GRCh38
rs771413152 Chr.16: 10440468-10440468 on Build GRCh38
rs377556995 Chr.16: 10440399-10440399 on Build GRCh38
rs192018149 Chr.16: 10440430-10440430 on Build GRCh38
rs546276724 Chr.16: 10440455-10440455 on Build GRCh38
rs374355408 Chr.16: 10440501-10440501 on Build GRCh38

View Full Product Details