Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 122934374-122934617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf63
Entrez Gene ID
79864
Gene Name
chromosome 11 open reading frame 63
Gene Aliases
-
UniGene
Hs.164705
Gene Chromosome Location
Chr.11: 122881640-122963862 on Build GRCh38
Transcripts
NM_024806.3 XM_005271680.4 XM_006718915.2 XM_011543009.2 XM_017018338.1 XM_017018339.1 XM_017018340.1 XM_017018341.1 XM_017018342.1 XM_017018343.1 XM_017018344.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1973493 c.1025A>T p.N342I Chr.11: 122934466-122934466 on Build GRCh38
1973496 c.1142C>T p.T381M Chr.11: 122934583-122934583 on Build GRCh38
1973491 c.996G>T p.W332C Chr.11: 122934437-122934437 on Build GRCh38
3808618 c.980A>T p.N327I Chr.11: 122934421-122934421 on Build GRCh38
4165675 c.1019C>A p.S340* Chr.11: 122934460-122934460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758821455 Chr.11: 122934537-122934537 on Build GRCh38
rs763609274 Chr.11: 122934487-122934487 on Build GRCh38
rs200668253 Chr.11: 122934583-122934583 on Build GRCh38
rs894946547 Chr.11: 122934433-122934433 on Build GRCh38
rs575543159 Chr.11: 122934466-122934466 on Build GRCh38
rs746310997 Chr.11: 122934530-122934530 on Build GRCh38
rs758165105 Chr.11: 122934426-122934426 on Build GRCh38
rs372643545 Chr.11: 122934547-122934547 on Build GRCh38
rs760379939 Chr.11: 122934565-122934565 on Build GRCh38
rs763703257 Chr.11: 122934581-122934581 on Build GRCh38
rs774372446 Chr.11: 122934550-122934550 on Build GRCh38
rs935110901 Chr.11: 122934451-122934451 on Build GRCh38
rs755697605 Chr.11: 122934482-122934482 on Build GRCh38
rs766338423 Chr.11: 122934412-122934412 on Build GRCh38
rs1046684947 Chr.11: 122934448-122934448 on Build GRCh38
rs753335141 Chr.11: 122934502-122934502 on Build GRCh38
rs914915676 Chr.11: 122934467-122934467 on Build GRCh38
rs780495678 Chr.11: 122934540-122934540 on Build GRCh38
rs759356744 Chr.11: 122934461-122934461 on Build GRCh38
rs150680420 Chr.11: 122934541-122934541 on Build GRCh38
rs779452627 Chr.11: 122934522-122934522 on Build GRCh38
rs75858776 Chr.11: 122934509-122934509 on Build GRCh38
rs1043340496 Chr.11: 122934520-122934520 on Build GRCh38
rs767350303 Chr.11: 122934556-122934556 on Build GRCh38
rs767172782 Chr.11: 122934465-122934465 on Build GRCh38
rs750940156 Chr.11: 122934590-122934590 on Build GRCh38
rs201240481 Chr.11: 122934548-122934548 on Build GRCh38
rs200529319 Chr.11: 122934558-122934558 on Build GRCh38
rs182156670 Chr.11: 122934553-122934553 on Build GRCh38
rs949258932 Chr.11: 122934443-122934443 on Build GRCh38
rs776919948 Chr.11: 122934545-122934545 on Build GRCh38
rs764612693 Chr.11: 122934584-122934584 on Build GRCh38
rs774056827 Chr.11: 122934459-122934459 on Build GRCh38

View Full Product Details