Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116760189-116760458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2
Entrez Gene ID
914
Gene Name
CD2 molecule
Gene Aliases
LFA-2, SRBC, T11
UniGene
Hs.523500
Gene Chromosome Location
Chr.1: 116754464-116769229 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4493049 c.408C>T p.S136S Chr.1: 116760427-116760427 on Build GRCh38
131063 c.427A>T p.T143S Chr.1: 116760446-116760446 on Build GRCh38
1747665 c.392C>G p.S131* Chr.1: 116760411-116760411 on Build GRCh38
894442 c.402G>T p.K134N Chr.1: 116760421-116760421 on Build GRCh38
3704867 c.389T>C p.V130A Chr.1: 116760408-116760408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768948538 Chr.1: 116760358-116760358 on Build GRCh38
rs773388806 Chr.1: 116760380-116760380 on Build GRCh38
rs1041606329 Chr.1: 116760238-116760238 on Build GRCh38
rs774555703 Chr.1: 116760360-116760360 on Build GRCh38
rs187014185 Chr.1: 116760318-116760318 on Build GRCh38
rs777272827 Chr.1: 116760431-116760431 on Build GRCh38
rs1048533969 Chr.1: 116760245-116760246 on Build GRCh38
rs910520833 Chr.1: 116760357-116760357 on Build GRCh38
rs1045092942 Chr.1: 116760306-116760306 on Build GRCh38
rs539458641 Chr.1: 116760253-116760253 on Build GRCh38
rs756744201 Chr.1: 116760435-116760435 on Build GRCh38
rs1003498571 Chr.1: 116760373-116760373 on Build GRCh38
rs762054502 Chr.1: 116760364-116760364 on Build GRCh38
rs760638704 Chr.1: 116760397-116760397 on Build GRCh38
rs774687074 Chr.1: 116760237-116760237 on Build GRCh38
rs563583333 Chr.1: 116760432-116760432 on Build GRCh38
rs575520883 Chr.1: 116760236-116760236 on Build GRCh38
rs780489954 Chr.1: 116760352-116760353 on Build GRCh38
rs191435301 Chr.1: 116760325-116760325 on Build GRCh38
rs376823879 Chr.1: 116760403-116760403 on Build GRCh38
rs138543209 Chr.1: 116760282-116760282 on Build GRCh38
rs1022535752 Chr.1: 116760295-116760295 on Build GRCh38
rs918795780 Chr.1: 116760240-116760240 on Build GRCh38
rs747381720 Chr.1: 116760391-116760391 on Build GRCh38
rs986532416 Chr.1: 116760301-116760301 on Build GRCh38
rs753814547 Chr.1: 116760416-116760416 on Build GRCh38
rs565087711 Chr.1: 116760243-116760243 on Build GRCh38
rs542035903 Chr.1: 116760424-116760424 on Build GRCh38
rs930038148 Chr.1: 116760244-116760245 on Build GRCh38
rs143587533 Chr.1: 116760366-116760366 on Build GRCh38
rs752328008 Chr.1: 116760426-116760426 on Build GRCh38
rs758023273 Chr.1: 116760427-116760427 on Build GRCh38
rs754835293 Chr.1: 116760421-116760421 on Build GRCh38
rs768911017 Chr.1: 116760394-116760395 on Build GRCh38
rs149317456 Chr.1: 116760296-116760296 on Build GRCh38
rs888545535 Chr.1: 116760286-116760286 on Build GRCh38
rs766492486 Chr.1: 116760405-116760405 on Build GRCh38

View Full Product Details