Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144545203-144545432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP39
Entrez Gene ID
80728
Gene Name
Rho GTPase activating protein 39
Gene Aliases
CrGAP, Vilse
UniGene
Hs.148988
Gene Chromosome Location
Chr.8: 144529179-144700520 on Build GRCh38
Transcripts
NM_001308207.1 NM_001308208.1 NM_025251.2 XM_011517308.1 XM_011517309.1 XM_011517312.2 XM_017013870.1 XM_017013871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2872028 c.2389C>T p.R797C Chr.8: 144545381-144545381 on Build GRCh38
334834 c.2514C>T p.D838D Chr.8: 144545256-144545256 on Build GRCh38
6603594 c.2407C>T p.R803C Chr.8: 144545363-144545363 on Build GRCh38
2872029 c.2379C>T p.T793T Chr.8: 144545391-144545391 on Build GRCh38
4480924 c.2469C>T p.S823S Chr.8: 144545301-144545301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760479420 Chr.8: 144545361-144545361 on Build GRCh38
rs888942635 Chr.8: 144545239-144545239 on Build GRCh38
rs755123319 Chr.8: 144545258-144545258 on Build GRCh38
rs764464947 Chr.8: 144545376-144545376 on Build GRCh38
rs758497562 Chr.8: 144545364-144545364 on Build GRCh38
rs868140624 Chr.8: 144545359-144545359 on Build GRCh38
rs773336066 Chr.8: 144545274-144545274 on Build GRCh38
rs752652890 Chr.8: 144545362-144545362 on Build GRCh38
rs778282421 Chr.8: 144545270-144545270 on Build GRCh38
rs751806798 Chr.8: 144545380-144545380 on Build GRCh38
rs148092254 Chr.8: 144545260-144545260 on Build GRCh38
rs750339896 Chr.8: 144545248-144545248 on Build GRCh38
rs767449644 Chr.8: 144545358-144545358 on Build GRCh38
rs769372274 Chr.8: 144545319-144545319 on Build GRCh38
rs761593712 Chr.8: 144545236-144545236 on Build GRCh38
rs774809694 Chr.8: 144545320-144545320 on Build GRCh38
rs767502642 Chr.8: 144545242-144545242 on Build GRCh38
rs200810387 Chr.8: 144545325-144545325 on Build GRCh38
rs781651492 Chr.8: 144545382-144545382 on Build GRCh38
rs773980215 Chr.8: 144545331-144545331 on Build GRCh38
rs138829800 Chr.8: 144545391-144545391 on Build GRCh38
rs758491166 Chr.8: 144545266-144545266 on Build GRCh38
rs781776360 Chr.8: 144545289-144545289 on Build GRCh38
rs144833316 Chr.8: 144545235-144545235 on Build GRCh38
rs139776939 Chr.8: 144545264-144545264 on Build GRCh38
rs763967522 Chr.8: 144545234-144545234 on Build GRCh38
rs773162144 Chr.8: 144545360-144545360 on Build GRCh38
rs762467776 Chr.8: 144545226-144545226 on Build GRCh38
rs757729796 Chr.8: 144545381-144545381 on Build GRCh38
rs61748959 Chr.8: 144545316-144545316 on Build GRCh38
rs149415890 Chr.8: 144545222-144545222 on Build GRCh38
rs775242688 Chr.8: 144545224-144545224 on Build GRCh38
rs767271501 Chr.8: 144545342-144545342 on Build GRCh38
rs993549396 Chr.8: 144545278-144545278 on Build GRCh38
rs757680596 Chr.8: 144545286-144545286 on Build GRCh38
rs761646995 Chr.8: 144545332-144545332 on Build GRCh38

View Full Product Details