Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89185323-89185584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD2
Entrez Gene ID
11057
Gene Name
abhydrolase domain containing 2
Gene Aliases
HS1-2, LABH2, PHPS1-2
UniGene
Hs.122337
Gene Chromosome Location
Chr.15: 89088150-89202360 on Build GRCh38
Transcripts
NM_007011.7 NM_152924.4 XM_005254832.1 XM_005254833.1 XM_011521165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244017 c.759G>T p.R253R Chr.15: 89185460-89185460 on Build GRCh38
396929 c.751C>T p.Q251* Chr.15: 89185452-89185452 on Build GRCh38
6143176 c.798G>C p.K266N Chr.15: 89185499-89185499 on Build GRCh38
4445272 c.723-3C>T p.? Chr.15: 89185421-89185421 on Build GRCh38
1665323 c.792_794delGAA p.K266delK Chr.15: 89185493-89185495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777652392 Chr.15: 89185545-89185545 on Build GRCh38
rs762986845 Chr.15: 89185381-89185382 on Build GRCh38
rs376550242 Chr.15: 89185531-89185531 on Build GRCh38
rs367650390 Chr.15: 89185417-89185417 on Build GRCh38
rs145464697 Chr.15: 89185445-89185445 on Build GRCh38
rs758663835 Chr.15: 89185540-89185540 on Build GRCh38
rs372134608 Chr.15: 89185392-89185392 on Build GRCh38
rs766672605 Chr.15: 89185408-89185408 on Build GRCh38
rs758370032 Chr.15: 89185421-89185421 on Build GRCh38
rs964844094 Chr.15: 89185547-89185547 on Build GRCh38
rs765666419 Chr.15: 89185536-89185536 on Build GRCh38
rs749058520 Chr.15: 89185558-89185558 on Build GRCh38
rs751759512 Chr.15: 89185546-89185546 on Build GRCh38
rs185898664 Chr.15: 89185382-89185382 on Build GRCh38
rs371827987 Chr.15: 89185458-89185458 on Build GRCh38
rs766170270 Chr.15: 89185543-89185562 on Build GRCh38
rs773895717 Chr.15: 89185386-89185386 on Build GRCh38
rs756635965 Chr.15: 89185407-89185407 on Build GRCh38
rs773911562 Chr.15: 89185510-89185510 on Build GRCh38
rs994926397 Chr.15: 89185550-89185550 on Build GRCh38
rs752814335 Chr.15: 89185539-89185539 on Build GRCh38
rs374726986 Chr.15: 89185411-89185411 on Build GRCh38
rs1047993112 Chr.15: 89185471-89185471 on Build GRCh38
rs921050688 Chr.15: 89185388-89185389 on Build GRCh38
rs548424325 Chr.15: 89185365-89185365 on Build GRCh38
rs1002249408 Chr.15: 89185525-89185525 on Build GRCh38
rs547465260 Chr.15: 89185373-89185373 on Build GRCh38
rs749149742 Chr.15: 89185505-89185505 on Build GRCh38
rs202074494 Chr.15: 89185511-89185511 on Build GRCh38
rs199736172 Chr.15: 89185553-89185553 on Build GRCh38
rs775442672 Chr.15: 89185375-89185375 on Build GRCh38
rs779053963 Chr.15: 89185412-89185412 on Build GRCh38
rs993875950 Chr.15: 89185463-89185463 on Build GRCh38
rs771391249 Chr.15: 89185552-89185552 on Build GRCh38
rs752792528 Chr.15: 89185414-89185414 on Build GRCh38
rs899130626 Chr.15: 89185543-89185543 on Build GRCh38
rs17851730 Chr.15: 89185459-89185459 on Build GRCh38
rs370112607 Chr.15: 89185530-89185530 on Build GRCh38
rs772792345 Chr.15: 89185515-89185515 on Build GRCh38
rs768033465 Chr.15: 89185391-89185391 on Build GRCh38
rs200800145 Chr.15: 89185390-89185390 on Build GRCh38
rs77592008 Chr.15: 89185464-89185464 on Build GRCh38
rs771433362 Chr.15: 89185514-89185514 on Build GRCh38
rs926437231 Chr.15: 89185389-89185389 on Build GRCh38
rs754204412 Chr.15: 89185409-89185409 on Build GRCh38
rs1006458302 Chr.15: 89185518-89185518 on Build GRCh38
rs779994462 Chr.15: 89185486-89185486 on Build GRCh38

View Full Product Details