Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15852149-15852408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1S2
Entrez Gene ID
8905
Gene Name
adaptor related protein complex 1 sigma 2 subunit
Gene Aliases
DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B
UniGene
Hs.121592
Gene Chromosome Location
Chr.X: 15825806-15855014 on Build GRCh38
Transcripts
NM_001272071.1 NM_003916.4 XM_005274614.3 XM_011545599.1 XM_017029925.1 XM_017029926.1

SNP Detail

SNP ID Chromosome Location
rs767055780 Chr.X: 15852375-15852375 on Build GRCh38
rs1006994319 Chr.X: 15852356-15852356 on Build GRCh38
rs759073386 Chr.X: 15852361-15852361 on Build GRCh38
rs909142817 Chr.X: 15852190-15852190 on Build GRCh38
rs749708694 Chr.X: 15852300-15852300 on Build GRCh38
rs376525848 Chr.X: 15852378-15852378 on Build GRCh38
rs971733313 Chr.X: 15852286-15852286 on Build GRCh38
rs1026822231 Chr.X: 15852321-15852321 on Build GRCh38
rs774166253 Chr.X: 15852329-15852329 on Build GRCh38
rs896168640 Chr.X: 15852182-15852182 on Build GRCh38
rs889589992 Chr.X: 15852334-15852334 on Build GRCh38
rs963012632 Chr.X: 15852208-15852208 on Build GRCh38
rs745487408 Chr.X: 15852335-15852335 on Build GRCh38
rs770683935 Chr.X: 15852328-15852328 on Build GRCh38
rs986089853 Chr.X: 15852173-15852173 on Build GRCh38
rs187909844 Chr.X: 15852293-15852293 on Build GRCh38
rs897066499 Chr.X: 15852231-15852231 on Build GRCh38
rs755826541 Chr.X: 15852240-15852240 on Build GRCh38
rs995378808 Chr.X: 15852276-15852276 on Build GRCh38
rs918968414 Chr.X: 15852376-15852376 on Build GRCh38
rs104894735 Chr.X: 15852371-15852371 on Build GRCh38

View Full Product Details