Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66579691-66579918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM2
Entrez Gene ID
146225
Gene Name
CKLF like MARVEL transmembrane domain containing 2
Gene Aliases
CKLFSF2
UniGene
Hs.195685
Gene Chromosome Location
Chr.16: 66579448-66588275 on Build GRCh38
Transcripts
NM_001199317.1 NM_144673.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3510814 c.181G>A p.E61K Chr.16: 66579788-66579788 on Build GRCh38
6144997 c.105G>A p.S35S Chr.16: 66579712-66579712 on Build GRCh38
4481652 c.252C>T p.L84L Chr.16: 66579859-66579859 on Build GRCh38
6226952 c.91G>A p.G31S Chr.16: 66579698-66579698 on Build GRCh38
5851177 c.203G>A p.C68Y Chr.16: 66579810-66579810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146637368 Chr.16: 66579864-66579864 on Build GRCh38
rs370543135 Chr.16: 66579773-66579773 on Build GRCh38
rs781633308 Chr.16: 66579798-66579798 on Build GRCh38
rs778989765 Chr.16: 66579818-66579818 on Build GRCh38
rs756562089 Chr.16: 66579805-66579805 on Build GRCh38
rs767256157 Chr.16: 66579887-66579887 on Build GRCh38
rs776501789 Chr.16: 66579730-66579730 on Build GRCh38
rs200227017 Chr.16: 66579772-66579772 on Build GRCh38
rs751207201 Chr.16: 66579779-66579779 on Build GRCh38
rs887206753 Chr.16: 66579868-66579868 on Build GRCh38
rs761550400 Chr.16: 66579874-66579874 on Build GRCh38
rs750727871 Chr.16: 66579888-66579888 on Build GRCh38
rs762057808 Chr.16: 66579721-66579721 on Build GRCh38
rs765287841 Chr.16: 66579753-66579753 on Build GRCh38
rs746230824 Chr.16: 66579832-66579833 on Build GRCh38
rs537890429 Chr.16: 66579741-66579741 on Build GRCh38
rs903927625 Chr.16: 66579768-66579768 on Build GRCh38
rs770666480 Chr.16: 66579727-66579727 on Build GRCh38
rs914269289 Chr.16: 66579785-66579785 on Build GRCh38
rs769598007 Chr.16: 66579827-66579827 on Build GRCh38
rs748940720 Chr.16: 66579800-66579800 on Build GRCh38
rs774757992 Chr.16: 66579809-66579809 on Build GRCh38
rs9925443 Chr.16: 66579788-66579788 on Build GRCh38
rs575414210 Chr.16: 66579740-66579740 on Build GRCh38
rs112263070 Chr.16: 66579895-66579895 on Build GRCh38
rs1030920227 Chr.16: 66579787-66579787 on Build GRCh38
rs759434314 Chr.16: 66579825-66579825 on Build GRCh38
rs577701143 Chr.16: 66579816-66579816 on Build GRCh38
rs752135930 Chr.16: 66579771-66579771 on Build GRCh38
rs371415575 Chr.16: 66579799-66579799 on Build GRCh38
rs775457714 Chr.16: 66579848-66579848 on Build GRCh38
rs768840389 Chr.16: 66579711-66579711 on Build GRCh38
rs187643040 Chr.16: 66579712-66579712 on Build GRCh38
rs368665656 Chr.16: 66579813-66579813 on Build GRCh38
rs759326774 Chr.16: 66579745-66579745 on Build GRCh38
rs751117468 Chr.16: 66579869-66579869 on Build GRCh38
rs111592429 Chr.16: 66579821-66579821 on Build GRCh38
rs772287237 Chr.16: 66579822-66579822 on Build GRCh38
rs1013433548 Chr.16: 66579801-66579801 on Build GRCh38
rs934164394 Chr.16: 66579847-66579848 on Build GRCh38
rs151139084 Chr.16: 66579775-66579775 on Build GRCh38
rs932122144 Chr.16: 66579728-66579728 on Build GRCh38
rs771408245 Chr.16: 66579784-66579794 on Build GRCh38

View Full Product Details