Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30007240-30007513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2A
Entrez Gene ID
8448
Gene Name
double C2 domain alpha
Gene Aliases
Doc2
UniGene
Hs.355281
Gene Chromosome Location
Chr.16: 30005514-30023280 on Build GRCh38
Transcripts
NM_001282062.1 NM_001282063.1 NM_001282068.1 NM_003586.2 NR_104089.1 NR_104090.1 XM_011545975.1 XM_017023776.1 XM_017023777.1 XM_017023778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3888381 c.576G>A p.G192G Chr.16: 30007251-30007251 on Build GRCh38
6647914 c.531C>T p.I177I Chr.16: 30007296-30007296 on Build GRCh38
6855439 c.584G>A p.R195H Chr.16: 30007243-30007243 on Build GRCh38
3508710 c.577G>A p.E193K Chr.16: 30007250-30007250 on Build GRCh38
6647915 c.535G>A p.V179I Chr.16: 30007292-30007292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746804118 Chr.16: 30007337-30007337 on Build GRCh38
rs528116198 Chr.16: 30007481-30007481 on Build GRCh38
rs1031612341 Chr.16: 30007264-30007264 on Build GRCh38
rs12921753 Chr.16: 30007399-30007399 on Build GRCh38
rs367572265 Chr.16: 30007292-30007292 on Build GRCh38
rs569339218 Chr.16: 30007418-30007418 on Build GRCh38
rs768639487 Chr.16: 30007341-30007341 on Build GRCh38
rs749818602 Chr.16: 30007306-30007306 on Build GRCh38
rs566931599 Chr.16: 30007421-30007421 on Build GRCh38
rs772626075 Chr.16: 30007349-30007349 on Build GRCh38
rs761701447 Chr.16: 30007345-30007345 on Build GRCh38
rs763452493 Chr.16: 30007457-30007457 on Build GRCh38
rs894900223 Chr.16: 30007454-30007454 on Build GRCh38
rs778198086 Chr.16: 30007305-30007305 on Build GRCh38
rs748026208 Chr.16: 30007272-30007272 on Build GRCh38
rs200107144 Chr.16: 30007312-30007312 on Build GRCh38
rs546869318 Chr.16: 30007487-30007487 on Build GRCh38
rs368624440 Chr.16: 30007395-30007395 on Build GRCh38
rs139432976 Chr.16: 30007286-30007286 on Build GRCh38
rs1039180959 Chr.16: 30007400-30007400 on Build GRCh38
rs368609924 Chr.16: 30007320-30007320 on Build GRCh38
rs371808877 Chr.16: 30007295-30007295 on Build GRCh38
rs970209135 Chr.16: 30007300-30007300 on Build GRCh38
rs191060572 Chr.16: 30007267-30007267 on Build GRCh38
rs550855109 Chr.16: 30007379-30007379 on Build GRCh38
rs939038534 Chr.16: 30007428-30007428 on Build GRCh38
rs544510908 Chr.16: 30007327-30007327 on Build GRCh38
rs1054847350 Chr.16: 30007434-30007434 on Build GRCh38
rs548522305 Chr.16: 30007415-30007415 on Build GRCh38
rs764650169 Chr.16: 30007484-30007484 on Build GRCh38
rs776468857 Chr.16: 30007343-30007343 on Build GRCh38
rs376380460 Chr.16: 30007350-30007350 on Build GRCh38
rs778900736 Chr.16: 30007316-30007316 on Build GRCh38
rs569613368 Chr.16: 30007466-30007491 on Build GRCh38
rs764708447 Chr.16: 30007283-30007283 on Build GRCh38
rs150867571 Chr.16: 30007275-30007275 on Build GRCh38
rs372335414 Chr.16: 30007323-30007323 on Build GRCh38
rs199801773 Chr.16: 30007296-30007296 on Build GRCh38
rs765187667 Chr.16: 30007346-30007346 on Build GRCh38
rs766027895 Chr.16: 30007351-30007351 on Build GRCh38

View Full Product Details