Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99390861-99391116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC1B
Entrez Gene ID
10095
Gene Name
actin related protein 2/3 complex subunit 1B
Gene Aliases
ARC41, p40-ARC, p41-ARC
UniGene
Hs.489284
Gene Chromosome Location
Chr.7: 99374675-99394801 on Build GRCh38
Transcripts
NM_005720.3 XM_006715825.1 XM_006715826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5873509 c.622G>A p.V208I Chr.7: 99391014-99391014 on Build GRCh38
3382029 c.588C>T p.F196F Chr.7: 99390980-99390980 on Build GRCh38
453657 c.540G>A p.P180P Chr.7: 99390932-99390932 on Build GRCh38
2867091 c.676G>A p.V226I Chr.7: 99391068-99391068 on Build GRCh38
4414135 c.615A>G p.V205V Chr.7: 99391007-99391007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751741478 Chr.7: 99391058-99391058 on Build GRCh38
rs771858187 Chr.7: 99391088-99391088 on Build GRCh38
rs758941292 Chr.7: 99390943-99390943 on Build GRCh38
rs761496510 Chr.7: 99390972-99390973 on Build GRCh38
rs371760619 Chr.7: 99391014-99391014 on Build GRCh38
rs138962515 Chr.7: 99390991-99390991 on Build GRCh38
rs376219355 Chr.7: 99391031-99391031 on Build GRCh38
rs772554544 Chr.7: 99390898-99390898 on Build GRCh38
rs1021339212 Chr.7: 99390941-99390941 on Build GRCh38
rs769279045 Chr.7: 99390993-99390993 on Build GRCh38
rs150907842 Chr.7: 99391049-99391049 on Build GRCh38
rs185268486 Chr.7: 99390888-99390888 on Build GRCh38
rs1046660850 Chr.7: 99391093-99391093 on Build GRCh38
rs777491800 Chr.7: 99390996-99390996 on Build GRCh38
rs149569034 Chr.7: 99391086-99391086 on Build GRCh38
rs781731759 Chr.7: 99391065-99391065 on Build GRCh38
rs745414166 Chr.7: 99391087-99391087 on Build GRCh38
rs775108226 Chr.7: 99391032-99391032 on Build GRCh38
rs746717899 Chr.7: 99391099-99391099 on Build GRCh38
rs367918454 Chr.7: 99390989-99390989 on Build GRCh38
rs138310411 Chr.7: 99391067-99391067 on Build GRCh38
rs137947738 Chr.7: 99390938-99390938 on Build GRCh38
rs117071116 Chr.7: 99391040-99391040 on Build GRCh38
rs752587907 Chr.7: 99390933-99390933 on Build GRCh38
rs756502444 Chr.7: 99391068-99391068 on Build GRCh38
rs577425201 Chr.7: 99391016-99391016 on Build GRCh38
rs145131136 Chr.7: 99390981-99390981 on Build GRCh38
rs780643186 Chr.7: 99390975-99390975 on Build GRCh38
rs774275938 Chr.7: 99390917-99390917 on Build GRCh38
rs762469116 Chr.7: 99390902-99390902 on Build GRCh38
rs760543643 Chr.7: 99391039-99391039 on Build GRCh38
rs562395006 Chr.7: 99391011-99391011 on Build GRCh38
rs573544884 Chr.7: 99390928-99390928 on Build GRCh38
rs781780782 Chr.7: 99390955-99390955 on Build GRCh38
rs767189739 Chr.7: 99390985-99390985 on Build GRCh38
rs778176237 Chr.7: 99391071-99391071 on Build GRCh38
rs140888847 Chr.7: 99390980-99390980 on Build GRCh38
rs200703477 Chr.7: 99390905-99390905 on Build GRCh38
rs34820590 Chr.7: 99390998-99390998 on Build GRCh38
rs199528626 Chr.7: 99390944-99390944 on Build GRCh38
rs759537436 Chr.7: 99390931-99390931 on Build GRCh38
rs753117732 Chr.7: 99390932-99390932 on Build GRCh38
rs759445881 Chr.7: 99391020-99391020 on Build GRCh38
rs748924502 Chr.7: 99391074-99391074 on Build GRCh38
rs368940243 Chr.7: 99391085-99391085 on Build GRCh38
rs755396037 Chr.7: 99391059-99391059 on Build GRCh38
rs763336010 Chr.7: 99391041-99391041 on Build GRCh38
rs769523636 Chr.7: 99390889-99390889 on Build GRCh38
rs781670733 Chr.7: 99390887-99390887 on Build GRCh38

View Full Product Details