Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241860506-241860741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO1
Entrez Gene ID
9156
Gene Name
exonuclease 1
Gene Aliases
HEX1, hExoI
UniGene
Hs.498248
Gene Chromosome Location
Chr.1: 241847967-241889939 on Build GRCh38
Transcripts
NM_001319224.1 NM_003686.4 NM_006027.4 NM_130398.3 XM_006711840.2 XM_011544321.1 XM_011544322.1 XM_011544323.2 XM_011544324.2 XM_017002793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681987 c.798G>A p.T266T Chr.1: 241860558-241860558 on Build GRCh38
7245376 c.873C>G p.I291M Chr.1: 241860633-241860633 on Build GRCh38
310958 c.797C>T p.T266M Chr.1: 241860557-241860557 on Build GRCh38
2050276 c.937G>T p.A313S Chr.1: 241860697-241860697 on Build GRCh38
6657043 c.829C>T p.R277W Chr.1: 241860589-241860589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763363627 Chr.1: 241860668-241860668 on Build GRCh38
rs201509012 Chr.1: 241860569-241860569 on Build GRCh38
rs752774825 Chr.1: 241860682-241860682 on Build GRCh38
rs765472230 Chr.1: 241860574-241860574 on Build GRCh38
rs569695378 Chr.1: 241860702-241860702 on Build GRCh38
rs745732917 Chr.1: 241860558-241860558 on Build GRCh38
rs746983719 Chr.1: 241860647-241860647 on Build GRCh38
rs1002966274 Chr.1: 241860579-241860579 on Build GRCh38
rs772807629 Chr.1: 241860663-241860663 on Build GRCh38
rs766904903 Chr.1: 241860589-241860589 on Build GRCh38
rs149397534 Chr.1: 241860580-241860580 on Build GRCh38
rs4149910 Chr.1: 241860656-241860656 on Build GRCh38
rs781383854 Chr.1: 241860536-241860536 on Build GRCh38
rs767539399 Chr.1: 241860681-241860681 on Build GRCh38
rs201323654 Chr.1: 241860679-241860679 on Build GRCh38
rs150105848 Chr.1: 241860557-241860557 on Build GRCh38
rs776390406 Chr.1: 241860570-241860570 on Build GRCh38
rs745668030 Chr.1: 241860608-241860608 on Build GRCh38
rs577333879 Chr.1: 241860696-241860696 on Build GRCh38
rs143367336 Chr.1: 241860669-241860669 on Build GRCh38
rs772048875 Chr.1: 241860560-241860560 on Build GRCh38
rs758181788 Chr.1: 241860611-241860611 on Build GRCh38
rs4149909 Chr.1: 241860596-241860596 on Build GRCh38
rs146714333 Chr.1: 241860685-241860685 on Build GRCh38
rs746723520 Chr.1: 241860566-241860566 on Build GRCh38
rs1019889690 Chr.1: 241860687-241860687 on Build GRCh38
rs763302707 Chr.1: 241860582-241860582 on Build GRCh38
rs923733715 Chr.1: 241860705-241860705 on Build GRCh38
rs548499993 Chr.1: 241860553-241860553 on Build GRCh38
rs775632538 Chr.1: 241860565-241860565 on Build GRCh38
rs774812938 Chr.1: 241860673-241860673 on Build GRCh38
rs148709490 Chr.1: 241860600-241860600 on Build GRCh38
rs761587261 Chr.1: 241860573-241860573 on Build GRCh38
rs764359141 Chr.1: 241860690-241860690 on Build GRCh38
rs905819666 Chr.1: 241860575-241860575 on Build GRCh38
rs780117642 Chr.1: 241860615-241860615 on Build GRCh38
rs368542606 Chr.1: 241860697-241860697 on Build GRCh38
rs769493108 Chr.1: 241860661-241860661 on Build GRCh38
rs999323643 Chr.1: 241860649-241860649 on Build GRCh38
rs748421983 Chr.1: 241860551-241860551 on Build GRCh38
rs554727186 Chr.1: 241860657-241860657 on Build GRCh38
rs747698568 Chr.1: 241860658-241860658 on Build GRCh38
rs200754492 Chr.1: 241860599-241860599 on Build GRCh38

View Full Product Details