Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52292092-52292317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYCTK
Entrez Gene ID
132158
Gene Name
glycerate kinase
Gene Aliases
HBEBP2, HBEBP4, HBeAgBP4A
UniGene
Hs.415312
Gene Chromosome Location
Chr.3: 52287558-52295256 on Build GRCh38
Transcripts
NM_001144951.1 NM_145262.3 NR_026699.1 NR_026700.1 NR_026701.1 NR_026702.1 XM_017005730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753300 c.720C>T p.I240I Chr.3: 52292274-52292274 on Build GRCh38
6164999 c.727G>A p.D243N Chr.3: 52292281-52292281 on Build GRCh38
5572114 c.763G>A p.G255S Chr.3: 52292317-52292317 on Build GRCh38
3595911 c.721C>T p.L241L Chr.3: 52292275-52292275 on Build GRCh38
5447572 c.740A>G p.D247G Chr.3: 52292294-52292294 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999460436 Chr.3: 52292213-52292213 on Build GRCh38
rs1012034922 Chr.3: 52292163-52292163 on Build GRCh38
rs75198060 Chr.3: 52292177-52292177 on Build GRCh38
rs958066187 Chr.3: 52292238-52292238 on Build GRCh38
rs764622289 Chr.3: 52292221-52292221 on Build GRCh38
rs573888726 Chr.3: 52292161-52292161 on Build GRCh38
rs540349478 Chr.3: 52292122-52292122 on Build GRCh38
rs762327679 Chr.3: 52292242-52292242 on Build GRCh38
rs374124131 Chr.3: 52292251-52292251 on Build GRCh38
rs534977554 Chr.3: 52292139-52292139 on Build GRCh38
rs367737334 Chr.3: 52292121-52292121 on Build GRCh38
rs186061935 Chr.3: 52292192-52292192 on Build GRCh38
rs1057180010 Chr.3: 52292151-52292151 on Build GRCh38
rs201846267 Chr.3: 52292190-52292190 on Build GRCh38
rs76507415 Chr.3: 52292178-52292178 on Build GRCh38
rs753350229 Chr.3: 52292285-52292285 on Build GRCh38
rs763382716 Chr.3: 52292215-52292215 on Build GRCh38
rs371261626 Chr.3: 52292181-52292181 on Build GRCh38
rs1031395719 Chr.3: 52292214-52292214 on Build GRCh38
rs553960662 Chr.3: 52292140-52292140 on Build GRCh38
rs77133273 Chr.3: 52292179-52292179 on Build GRCh38
rs774990271 Chr.3: 52292227-52292227 on Build GRCh38
rs1037423162 Chr.3: 52292159-52292159 on Build GRCh38
rs989530824 Chr.3: 52292244-52292244 on Build GRCh38

View Full Product Details