Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65887927-65888194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBP
Entrez Gene ID
9158
Gene Name
FGF1 intracellular binding protein
Gene Aliases
FGFIBP, FIBP-1
UniGene
Hs.7768
Gene Chromosome Location
Chr.11: 65883740-65888539 on Build GRCh38
Transcripts
NM_004214.4 NM_198897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5562901 c.219G>A p.P73P Chr.11: 65887999-65887999 on Build GRCh38
3666972 c.117G>T p.S39S Chr.11: 65888101-65888101 on Build GRCh38
4035583 c.268G>A p.A90T Chr.11: 65887950-65887950 on Build GRCh38
6134241 c.247C>A p.Q83K Chr.11: 65887971-65887971 on Build GRCh38
5038948 c.268G>T p.A90S Chr.11: 65887950-65887950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747780320 Chr.11: 65888101-65888101 on Build GRCh38
rs767828364 Chr.11: 65888006-65888006 on Build GRCh38
rs556290751 Chr.11: 65888077-65888077 on Build GRCh38
rs765532458 Chr.11: 65888069-65888069 on Build GRCh38
rs766679393 Chr.11: 65887950-65887950 on Build GRCh38
rs140745794 Chr.11: 65888014-65888014 on Build GRCh38
rs754811803 Chr.11: 65888096-65888096 on Build GRCh38
rs771515201 Chr.11: 65888056-65888056 on Build GRCh38
rs777324318 Chr.11: 65888113-65888113 on Build GRCh38
rs746203212 Chr.11: 65888124-65888124 on Build GRCh38
rs145313106 Chr.11: 65887969-65887969 on Build GRCh38
rs1037073520 Chr.11: 65887961-65887961 on Build GRCh38
rs773275673 Chr.11: 65888042-65888042 on Build GRCh38
rs138356472 Chr.11: 65888078-65888078 on Build GRCh38
rs770311964 Chr.11: 65888125-65888125 on Build GRCh38
rs34255955 Chr.11: 65888068-65888068 on Build GRCh38
rs898067695 Chr.11: 65888061-65888061 on Build GRCh38
rs556176162 Chr.11: 65887990-65887990 on Build GRCh38
rs955872027 Chr.11: 65888146-65888146 on Build GRCh38
rs536278662 Chr.11: 65887972-65887972 on Build GRCh38
rs371916142 Chr.11: 65888092-65888092 on Build GRCh38
rs750309970 Chr.11: 65888088-65888088 on Build GRCh38
rs540588065 Chr.11: 65888166-65888166 on Build GRCh38
rs886037928 Chr.11: 65888042-65888043 on Build GRCh38
rs756003463 Chr.11: 65888089-65888089 on Build GRCh38
rs777126480 Chr.11: 65888064-65888064 on Build GRCh38
rs757870004 Chr.11: 65888106-65888106 on Build GRCh38
rs778501484 Chr.11: 65888099-65888099 on Build GRCh38
rs11559155 Chr.11: 65888001-65888001 on Build GRCh38
rs775894419 Chr.11: 65888075-65888075 on Build GRCh38
rs970349216 Chr.11: 65888121-65888121 on Build GRCh38
rs777079373 Chr.11: 65888085-65888086 on Build GRCh38
rs777150206 Chr.11: 65888109-65888109 on Build GRCh38
rs145310878 Chr.11: 65888095-65888095 on Build GRCh38
rs746594129 Chr.11: 65888112-65888112 on Build GRCh38
rs758456440 Chr.11: 65887963-65887963 on Build GRCh38
rs764101631 Chr.11: 65888086-65888086 on Build GRCh38
rs917177309 Chr.11: 65888147-65888147 on Build GRCh38
rs566724173 Chr.11: 65888173-65888173 on Build GRCh38
rs560814040 Chr.11: 65888167-65888167 on Build GRCh38
rs765617232 Chr.11: 65887960-65887960 on Build GRCh38
rs778589332 Chr.11: 65888016-65888016 on Build GRCh38
rs775804391 Chr.11: 65888160-65888160 on Build GRCh38
rs773722173 Chr.11: 65888059-65888059 on Build GRCh38
rs1007774881 Chr.11: 65888076-65888076 on Build GRCh38
rs1049819782 Chr.11: 65888021-65888021 on Build GRCh38
rs369931187 Chr.11: 65887979-65887979 on Build GRCh38
rs141319621 Chr.11: 65887956-65887956 on Build GRCh38
rs199621848 Chr.11: 65887962-65887962 on Build GRCh38
rs771257958 Chr.11: 65888062-65888062 on Build GRCh38
rs981845401 Chr.11: 65888142-65888142 on Build GRCh38
rs11559153 Chr.11: 65887999-65887999 on Build GRCh38
rs747630653 Chr.11: 65888029-65888029 on Build GRCh38
rs747481931 Chr.11: 65888060-65888060 on Build GRCh38
rs114777258 Chr.11: 65888083-65888083 on Build GRCh38

View Full Product Details