Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177405191-177405438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F12
Entrez Gene ID
2161
Gene Name
coagulation factor XII
Gene Aliases
HAE3, HAEX, HAF
UniGene
Hs.1321
Gene Chromosome Location
Chr.5: 177402138-177409576 on Build GRCh38
Transcripts
NM_000505.3 XM_011534462.2 XM_017009773.1

Gene Symbol
SLC34A1
Entrez Gene ID
6569
Gene Name
solute carrier family 34 member 1
Gene Aliases
FRTS2, HCINF2, NAPI-3, NPHLOP1, NPT2, NPTIIa, SLC11, SLC17A2
UniGene
Hs.936
Gene Chromosome Location
Chr.5: 177384431-177412021 on Build GRCh38
Transcripts
NM_000505.3 XM_011534462.2 XM_017009773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6361740 c.304A>G p.S102G Chr.5: 177405416-177405416 on Build GRCh38
4833017 c.360C>T p.L120L Chr.5: 177405360-177405360 on Build GRCh38
5427892 c.368A>C p.Q123P Chr.5: 177405352-177405352 on Build GRCh38
3366005 c.332T>G p.V111G Chr.5: 177405388-177405388 on Build GRCh38
5737439 c.389G>T p.C130F Chr.5: 177405331-177405331 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs149718961 Chr.5: 177405241-177405241 on Build GRCh38
rs767336155 Chr.5: 177405301-177405301 on Build GRCh38
rs754877167 Chr.5: 177405374-177405374 on Build GRCh38
rs558195458 Chr.5: 177405310-177405310 on Build GRCh38
rs796411771 Chr.5: 177405337-177405337 on Build GRCh38
rs747300174 Chr.5: 177405416-177405417 on Build GRCh38
rs982008123 Chr.5: 177405285-177405285 on Build GRCh38
rs753022412 Chr.5: 177405315-177405317 on Build GRCh38
rs201422427 Chr.5: 177405389-177405389 on Build GRCh38
rs755147039 Chr.5: 177405214-177405214 on Build GRCh38
rs73336290 Chr.5: 177405294-177405294 on Build GRCh38
rs577869365 Chr.5: 177405240-177405240 on Build GRCh38
rs776217044 Chr.5: 177405224-177405224 on Build GRCh38
rs960620418 Chr.5: 177405290-177405290 on Build GRCh38
rs758487283 Chr.5: 177405368-177405368 on Build GRCh38
rs761533070 Chr.5: 177405231-177405231 on Build GRCh38
rs778949627 Chr.5: 177405351-177405351 on Build GRCh38
rs56022250 Chr.5: 177405258-177405258 on Build GRCh38
rs373624367 Chr.5: 177405298-177405298 on Build GRCh38
rs769220140 Chr.5: 177405375-177405375 on Build GRCh38
rs757509766 Chr.5: 177405313-177405313 on Build GRCh38
rs778724407 Chr.5: 177405380-177405380 on Build GRCh38
rs865792091 Chr.5: 177405330-177405330 on Build GRCh38
rs940647627 Chr.5: 177405297-177405297 on Build GRCh38
rs764741084 Chr.5: 177405272-177405272 on Build GRCh38
rs778208227 Chr.5: 177405411-177405411 on Build GRCh38
rs956630674 Chr.5: 177405238-177405238 on Build GRCh38
rs769517704 Chr.5: 177405412-177405412 on Build GRCh38
rs942858184 Chr.5: 177405401-177405401 on Build GRCh38
rs765400848 Chr.5: 177405282-177405282 on Build GRCh38
rs772800879 Chr.5: 177405281-177405281 on Build GRCh38
rs745604326 Chr.5: 177405219-177405219 on Build GRCh38
rs41309750 Chr.5: 177405361-177405361 on Build GRCh38
rs140243617 Chr.5: 177405372-177405372 on Build GRCh38
rs969203373 Chr.5: 177405278-177405278 on Build GRCh38
rs564511690 Chr.5: 177405245-177405245 on Build GRCh38
rs746002981 Chr.5: 177405358-177405358 on Build GRCh38
rs377676479 Chr.5: 177405392-177405392 on Build GRCh38
rs769132100 Chr.5: 177405214-177405214 on Build GRCh38
rs775219086 Chr.5: 177405251-177405251 on Build GRCh38
rs1023391513 Chr.5: 177405263-177405263 on Build GRCh38

View Full Product Details