Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27638103-27638358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPN1
Entrez Gene ID
11321
Gene Name
GPN-loop GTPase 1
Gene Aliases
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1
UniGene
Hs.18259
Gene Chromosome Location
Chr.2: 27628648-27650846 on Build GRCh38
Transcripts
NM_001145047.1 NM_001145048.1 NM_001145049.1 NM_007266.3 NR_026735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4093656 c.532T>A p.Y178N Chr.2: 27638217-27638217 on Build GRCh38
721341 c.528C>T p.I176I Chr.2: 27638213-27638213 on Build GRCh38
2940625 c.562A>C p.M188L Chr.2: 27638247-27638247 on Build GRCh38
6380406 c.547C>T p.P183S Chr.2: 27638232-27638232 on Build GRCh38
1019849 c.551T>C p.F184S Chr.2: 27638236-27638236 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758566833 Chr.2: 27638179-27638179 on Build GRCh38
rs769311274 Chr.2: 27638219-27638219 on Build GRCh38
rs879103840 Chr.2: 27638172-27638172 on Build GRCh38
rs775994531 Chr.2: 27638250-27638250 on Build GRCh38
rs529675833 Chr.2: 27638300-27638300 on Build GRCh38
rs751336566 Chr.2: 27638306-27638306 on Build GRCh38
rs750617782 Chr.2: 27638171-27638171 on Build GRCh38
rs762259028 Chr.2: 27638271-27638271 on Build GRCh38
rs142201674 Chr.2: 27638309-27638309 on Build GRCh38
rs767960859 Chr.2: 27638324-27638324 on Build GRCh38
rs892283267 Chr.2: 27638244-27638244 on Build GRCh38
rs1050047834 Chr.2: 27638137-27638137 on Build GRCh38
rs375118761 Chr.2: 27638184-27638184 on Build GRCh38
rs879453949 Chr.2: 27638140-27638140 on Build GRCh38
rs768202357 Chr.2: 27638152-27638152 on Build GRCh38
rs1022227999 Chr.2: 27638278-27638278 on Build GRCh38
rs749062466 Chr.2: 27638164-27638164 on Build GRCh38
rs1052304969 Chr.2: 27638208-27638208 on Build GRCh38
rs747582566 Chr.2: 27638217-27638217 on Build GRCh38
rs748913982 Chr.2: 27638227-27638227 on Build GRCh38
rs770712926 Chr.2: 27638247-27638247 on Build GRCh38
rs534243286 Chr.2: 27638191-27638191 on Build GRCh38
rs202179483 Chr.2: 27638185-27638185 on Build GRCh38
rs773816404 Chr.2: 27638263-27638263 on Build GRCh38
rs781158994 Chr.2: 27638216-27638216 on Build GRCh38
rs754902082 Chr.2: 27638211-27638211 on Build GRCh38
rs368255706 Chr.2: 27638294-27638294 on Build GRCh38
rs769262937 Chr.2: 27638258-27638258 on Build GRCh38
rs959972645 Chr.2: 27638224-27638224 on Build GRCh38
rs748878379 Chr.2: 27638139-27638139 on Build GRCh38
rs1034279981 Chr.2: 27638325-27638325 on Build GRCh38
rs777170740 Chr.2: 27638222-27638222 on Build GRCh38
rs761178257 Chr.2: 27638256-27638256 on Build GRCh38
rs763093020 Chr.2: 27638295-27638295 on Build GRCh38
rs1036005171 Chr.2: 27638181-27638181 on Build GRCh38
rs765308292 Chr.2: 27638273-27638273 on Build GRCh38
rs1008642299 Chr.2: 27638207-27638207 on Build GRCh38

View Full Product Details