Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7074921-7075166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC10A
Entrez Gene ID
10462
Gene Name
C-type lectin domain family 10 member A
Gene Aliases
CD301, CLECSF13, CLECSF14, HML, HML2, MGL
UniGene
Hs.54403
Gene Chromosome Location
Chr.17: 7074537-7080635 on Build GRCh38
Transcripts
NM_006344.2 NM_182906.2 XM_005256412.1 XM_011523613.1 XM_011523614.2 XM_011523615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2153435 c.855C>T p.F285F Chr.17: 7075150-7075150 on Build GRCh38
983738 c.888C>A p.C296* Chr.17: 7075117-7075117 on Build GRCh38
1750280 c.914A>T p.E305V Chr.17: 7075091-7075091 on Build GRCh38
5782965 c.869G>T p.R290M Chr.17: 7075136-7075136 on Build GRCh38
6081586 c.933C>A p.T311T Chr.17: 7075072-7075072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769675310 Chr.17: 7074970-7074970 on Build GRCh38
rs763602748 Chr.17: 7075126-7075126 on Build GRCh38
rs777554621 Chr.17: 7075096-7075096 on Build GRCh38
rs200373971 Chr.17: 7075122-7075122 on Build GRCh38
rs149746467 Chr.17: 7075139-7075139 on Build GRCh38
rs772146117 Chr.17: 7075012-7075012 on Build GRCh38
rs376666860 Chr.17: 7075086-7075086 on Build GRCh38
rs773095830 Chr.17: 7075029-7075029 on Build GRCh38
rs752233615 Chr.17: 7075039-7075039 on Build GRCh38
rs532670931 Chr.17: 7075125-7075125 on Build GRCh38
rs748103007 Chr.17: 7075017-7075017 on Build GRCh38
rs750644413 Chr.17: 7075077-7075077 on Build GRCh38
rs759196587 Chr.17: 7075036-7075036 on Build GRCh38
rs764504573 Chr.17: 7075043-7075043 on Build GRCh38
rs867799020 Chr.17: 7074996-7074996 on Build GRCh38
rs113846982 Chr.17: 7074984-7074984 on Build GRCh38
rs386794994 Chr.17: 7075000-7075001 on Build GRCh38
rs774156162 Chr.17: 7075112-7075112 on Build GRCh38
rs538335075 Chr.17: 7075128-7075128 on Build GRCh38
rs188363668 Chr.17: 7074985-7074985 on Build GRCh38
rs774079173 Chr.17: 7075030-7075030 on Build GRCh38
rs962063260 Chr.17: 7075118-7075118 on Build GRCh38
rs576253599 Chr.17: 7074993-7074993 on Build GRCh38
rs560873499 Chr.17: 7075107-7075107 on Build GRCh38
rs758573050 Chr.17: 7075083-7075083 on Build GRCh38
rs906689242 Chr.17: 7075082-7075082 on Build GRCh38
rs754288455 Chr.17: 7075053-7075053 on Build GRCh38
rs761307050 Chr.17: 7075041-7075041 on Build GRCh38
rs138660752 Chr.17: 7074976-7074976 on Build GRCh38
rs552400469 Chr.17: 7075133-7075133 on Build GRCh38
rs770634151 Chr.17: 7075111-7075111 on Build GRCh38
rs962554995 Chr.17: 7075049-7075049 on Build GRCh38
rs139844327 Chr.17: 7075123-7075123 on Build GRCh38
rs935433730 Chr.17: 7075025-7075025 on Build GRCh38
rs372729049 Chr.17: 7075141-7075141 on Build GRCh38
rs746049589 Chr.17: 7075003-7075003 on Build GRCh38
rs371803977 Chr.17: 7075028-7075028 on Build GRCh38
rs375775825 Chr.17: 7075032-7075032 on Build GRCh38
rs991113331 Chr.17: 7075140-7075140 on Build GRCh38
rs762647211 Chr.17: 7075031-7075031 on Build GRCh38
rs201054202 Chr.17: 7075037-7075037 on Build GRCh38
rs371228002 Chr.17: 7075092-7075092 on Build GRCh38
rs146844702 Chr.17: 7075014-7075014 on Build GRCh38
rs774512949 Chr.17: 7075010-7075010 on Build GRCh38
rs766839218 Chr.17: 7074948-7074965 on Build GRCh38
rs191811013 Chr.17: 7074977-7074977 on Build GRCh38
rs367569967 Chr.17: 7075016-7075016 on Build GRCh38
rs546990321 Chr.17: 7075068-7075068 on Build GRCh38
rs15623 Chr.17: 7075000-7075000 on Build GRCh38
rs757579133 Chr.17: 7075059-7075059 on Build GRCh38
rs78425351 Chr.17: 7075001-7075001 on Build GRCh38
rs147465833 Chr.17: 7075093-7075093 on Build GRCh38

View Full Product Details