Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45793858-45794089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTTIP1
Entrez Gene ID
116092
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Gene Aliases
C20orf167, Tdif1, dJ447F3.4
UniGene
Hs.472852
Gene Chromosome Location
Chr.20: 45791937-45811427 on Build GRCh38
Transcripts
NM_052951.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1027279 c.188C>A p.P63H Chr.20: 45793932-45793932 on Build GRCh38
6863940 c.199A>G p.M67V Chr.20: 45793943-45793943 on Build GRCh38
1190268 c.270G>A p.M90I Chr.20: 45794014-45794014 on Build GRCh38
4434766 c.243G>C p.E81D Chr.20: 45793987-45793987 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs957710281 Chr.20: 45794059-45794061 on Build GRCh38
rs763369669 Chr.20: 45793956-45793956 on Build GRCh38
rs139834669 Chr.20: 45793961-45793961 on Build GRCh38
rs750176979 Chr.20: 45793897-45793897 on Build GRCh38
rs144493047 Chr.20: 45793981-45793981 on Build GRCh38
rs780193888 Chr.20: 45793990-45793990 on Build GRCh38
rs751807155 Chr.20: 45793991-45793991 on Build GRCh38
rs6032545 Chr.20: 45794066-45794066 on Build GRCh38
rs757308467 Chr.20: 45794064-45794064 on Build GRCh38
rs35492126 Chr.20: 45793987-45793987 on Build GRCh38
rs770973781 Chr.20: 45794060-45794060 on Build GRCh38
rs778310350 Chr.20: 45794047-45794047 on Build GRCh38
rs763728435 Chr.20: 45793975-45793975 on Build GRCh38
rs556797395 Chr.20: 45793982-45793982 on Build GRCh38
rs1000395850 Chr.20: 45794033-45794033 on Build GRCh38
rs1039138537 Chr.20: 45794021-45794021 on Build GRCh38
rs760354326 Chr.20: 45793964-45793964 on Build GRCh38
rs373752354 Chr.20: 45793893-45793893 on Build GRCh38
rs758609028 Chr.20: 45793903-45793903 on Build GRCh38
rs757222596 Chr.20: 45793892-45793892 on Build GRCh38
rs748654943 Chr.20: 45793937-45793937 on Build GRCh38
rs755123447 Chr.20: 45794005-45794005 on Build GRCh38
rs959746344 Chr.20: 45793896-45793896 on Build GRCh38
rs781260133 Chr.20: 45794012-45794012 on Build GRCh38
rs544617000 Chr.20: 45793913-45793913 on Build GRCh38
rs542614446 Chr.20: 45794053-45794053 on Build GRCh38
rs768366376 Chr.20: 45794067-45794067 on Build GRCh38
rs751419626 Chr.20: 45794061-45794061 on Build GRCh38
rs911984145 Chr.20: 45794057-45794057 on Build GRCh38
rs747135620 Chr.20: 45793908-45793908 on Build GRCh38
rs780321251 Chr.20: 45793904-45793904 on Build GRCh38
rs775260181 Chr.20: 45793963-45793963 on Build GRCh38
rs773504772 Chr.20: 45793955-45793955 on Build GRCh38
rs753849329 Chr.20: 45793889-45793889 on Build GRCh38
rs781204908 Chr.20: 45793923-45793923 on Build GRCh38
rs771348107 Chr.20: 45794058-45794058 on Build GRCh38
rs770269960 Chr.20: 45794027-45794027 on Build GRCh38
rs753332745 Chr.20: 45793980-45793980 on Build GRCh38
rs770274811 Chr.20: 45793951-45793951 on Build GRCh38
rs17352119 Chr.20: 45794014-45794014 on Build GRCh38

View Full Product Details