Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73498541-73498804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTF3
Entrez Gene ID
689
Gene Name
basic transcription factor 3
Gene Aliases
BETA-NAC, BTF3a, BTF3b, NACB
UniGene
Hs.591768
Gene Chromosome Location
Chr.5: 73498425-73505623 on Build GRCh38
Transcripts
NM_001037637.1 NM_001207.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4388456 c.66C>A p.C22* Chr.5: 73498733-73498733 on Build GRCh38
6416081 c.122A>T p.Q41L Chr.5: 73498789-73498789 on Build GRCh38
5433182 c.116G>A p.R39Q Chr.5: 73498783-73498783 on Build GRCh38
6241722 c.66C>T p.C22C Chr.5: 73498733-73498733 on Build GRCh38
6431095 c.91C>T p.Q31* Chr.5: 73498758-73498758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755340655 Chr.5: 73498675-73498675 on Build GRCh38
rs543140817 Chr.5: 73498668-73498668 on Build GRCh38
rs758924385 Chr.5: 73498696-73498696 on Build GRCh38
rs375502008 Chr.5: 73498700-73498700 on Build GRCh38
rs757317946 Chr.5: 73498734-73498734 on Build GRCh38
rs906717629 Chr.5: 73498732-73498732 on Build GRCh38
rs746927933 Chr.5: 73498705-73498705 on Build GRCh38
rs781134656 Chr.5: 73498650-73498652 on Build GRCh38
rs767504951 Chr.5: 73498656-73498656 on Build GRCh38
rs868180059 Chr.5: 73498678-73498678 on Build GRCh38
rs370228487 Chr.5: 73498716-73498716 on Build GRCh38
rs761827713 Chr.5: 73498653-73498653 on Build GRCh38
rs765666541 Chr.5: 73498685-73498685 on Build GRCh38
rs766190850 Chr.5: 73498661-73498661 on Build GRCh38
rs200658747 Chr.5: 73498618-73498618 on Build GRCh38
rs911238120 Chr.5: 73498767-73498767 on Build GRCh38
rs924035735 Chr.5: 73498643-73498643 on Build GRCh38
rs749097926 Chr.5: 73498632-73498632 on Build GRCh38
rs752955577 Chr.5: 73498690-73498690 on Build GRCh38
rs896915700 Chr.5: 73498774-73498774 on Build GRCh38
rs1036457953 Chr.5: 73498741-73498741 on Build GRCh38
rs768464646 Chr.5: 73498644-73498644 on Build GRCh38
rs978308105 Chr.5: 73498620-73498620 on Build GRCh38
rs563364749 Chr.5: 73498577-73498577 on Build GRCh38
rs753509680 Chr.5: 73498667-73498667 on Build GRCh38
rs760256228 Chr.5: 73498660-73498660 on Build GRCh38
rs955444008 Chr.5: 73498714-73498714 on Build GRCh38
rs1017497520 Chr.5: 73498580-73498580 on Build GRCh38
rs746130133 Chr.5: 73498738-73498738 on Build GRCh38
rs772853105 Chr.5: 73498657-73498657 on Build GRCh38
rs916583663 Chr.5: 73498694-73498694 on Build GRCh38
rs866966564 Chr.5: 73498692-73498692 on Build GRCh38
rs912416278 Chr.5: 73498573-73498573 on Build GRCh38
rs773973160 Chr.5: 73498650-73498650 on Build GRCh38
rs965225700 Chr.5: 73498584-73498584 on Build GRCh38

View Full Product Details