Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95201792-95202063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM6
Entrez Gene ID
142827
Gene Name
acyl-CoA synthetase medium-chain family member 6
Gene Aliases
C10orf129, bA310E22.3
UniGene
Hs.134229
Gene Chromosome Location
Chr.10: 95194200-95228929 on Build GRCh38
Transcripts
NM_207321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
229169 c.225G>A p.W75* Chr.10: 95202017-95202017 on Build GRCh38
5937419 c.196G>A p.G66R Chr.10: 95201988-95201988 on Build GRCh38
7357932 c.228G>C p.K76N Chr.10: 95202020-95202020 on Build GRCh38
398410 c.201C>T p.L67L Chr.10: 95201993-95201993 on Build GRCh38
1970968 c.222C>T p.L74L Chr.10: 95202014-95202014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981965671 Chr.10: 95201999-95201999 on Build GRCh38
rs971253733 Chr.10: 95201988-95201988 on Build GRCh38
rs867609011 Chr.10: 95202004-95202004 on Build GRCh38
rs201108997 Chr.10: 95202037-95202037 on Build GRCh38
rs977519484 Chr.10: 95201972-95201972 on Build GRCh38
rs991229115 Chr.10: 95201944-95201944 on Build GRCh38
rs182733896 Chr.10: 95201884-95201884 on Build GRCh38
rs909823672 Chr.10: 95202034-95202034 on Build GRCh38
rs921297291 Chr.10: 95202016-95202016 on Build GRCh38
rs1014866330 Chr.10: 95201823-95201823 on Build GRCh38
rs866586245 Chr.10: 95202017-95202017 on Build GRCh38
rs924864175 Chr.10: 95201979-95201979 on Build GRCh38
rs765703861 Chr.10: 95202005-95202005 on Build GRCh38
rs80081228 Chr.10: 95201975-95201975 on Build GRCh38
rs958598474 Chr.10: 95201913-95201913 on Build GRCh38
rs961956658 Chr.10: 95201841-95201841 on Build GRCh38
rs983975912 Chr.10: 95202021-95202021 on Build GRCh38
rs559069068 Chr.10: 95201940-95201940 on Build GRCh38
rs1021004272 Chr.10: 95201953-95201953 on Build GRCh38
rs112354911 Chr.10: 95202020-95202020 on Build GRCh38
rs995157836 Chr.10: 95201844-95201852 on Build GRCh38
rs1027364275 Chr.10: 95201859-95201859 on Build GRCh38
rs541264186 Chr.10: 95202009-95202009 on Build GRCh38
rs988549681 Chr.10: 95201902-95201902 on Build GRCh38
rs1032606566 Chr.10: 95201820-95201820 on Build GRCh38
rs936299659 Chr.10: 95201987-95201987 on Build GRCh38
rs549780713 Chr.10: 95201950-95201969 on Build GRCh38
rs750778872 Chr.10: 95202008-95202008 on Build GRCh38
rs971556933 Chr.10: 95201961-95201961 on Build GRCh38
rs958000449 Chr.10: 95201871-95201871 on Build GRCh38

View Full Product Details