Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48083852-48084107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC12
Entrez Gene ID
94160
Gene Name
ATP binding cassette subfamily C member 12
Gene Aliases
MRP9
UniGene
Hs.410111
Gene Chromosome Location
Chr.16: 48082973-48146770 on Build GRCh38
Transcripts
NM_033226.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1172333 c.3976G>T p.V1326F Chr.16: 48083926-48083926 on Build GRCh38
703680 c.3943C>A p.L1315I Chr.16: 48083959-48083959 on Build GRCh38
703678 c.3942C>A p.R1314R Chr.16: 48083960-48083960 on Build GRCh38
435291 c.3959A>T p.N1320I Chr.16: 48083943-48083943 on Build GRCh38
7332140 c.3897C>G p.I1299M Chr.16: 48084005-48084005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746612189 Chr.16: 48084054-48084054 on Build GRCh38
rs372109104 Chr.16: 48084071-48084071 on Build GRCh38
rs770490894 Chr.16: 48084068-48084068 on Build GRCh38
rs961612785 Chr.16: 48083942-48083942 on Build GRCh38
rs368257926 Chr.16: 48083926-48083926 on Build GRCh38
rs778075310 Chr.16: 48083904-48083904 on Build GRCh38
rs771665597 Chr.16: 48083938-48083938 on Build GRCh38
rs751803761 Chr.16: 48083997-48083997 on Build GRCh38
rs779629498 Chr.16: 48084023-48084023 on Build GRCh38
rs752772848 Chr.16: 48083988-48083988 on Build GRCh38
rs761090320 Chr.16: 48083930-48083931 on Build GRCh38
rs753807498 Chr.16: 48083886-48083886 on Build GRCh38
rs368282257 Chr.16: 48083982-48083982 on Build GRCh38
rs747241166 Chr.16: 48083905-48083905 on Build GRCh38
rs144766210 Chr.16: 48083996-48083996 on Build GRCh38
rs757480312 Chr.16: 48084013-48084013 on Build GRCh38
rs773783948 Chr.16: 48083927-48083927 on Build GRCh38
rs781478647 Chr.16: 48083913-48083913 on Build GRCh38
rs771541274 Chr.16: 48084049-48084049 on Build GRCh38
rs759479096 Chr.16: 48083981-48083981 on Build GRCh38
rs147483326 Chr.16: 48084032-48084032 on Build GRCh38
rs776303012 Chr.16: 48083969-48083969 on Build GRCh38
rs763052571 Chr.16: 48084081-48084081 on Build GRCh38
rs766181366 Chr.16: 48083968-48083968 on Build GRCh38
rs747689431 Chr.16: 48083934-48083934 on Build GRCh38
rs758527358 Chr.16: 48083995-48083995 on Build GRCh38
rs749846113 Chr.16: 48084043-48084043 on Build GRCh38
rs757612353 Chr.16: 48083909-48083909 on Build GRCh38
rs200251351 Chr.16: 48084050-48084050 on Build GRCh38
rs140699394 Chr.16: 48083961-48083961 on Build GRCh38
rs752824292 Chr.16: 48083895-48083895 on Build GRCh38
rs750114924 Chr.16: 48083896-48083896 on Build GRCh38
rs764222163 Chr.16: 48084083-48084083 on Build GRCh38
rs776446335 Chr.16: 48084069-48084069 on Build GRCh38
rs780564797 Chr.16: 48084038-48084038 on Build GRCh38
rs1028318850 Chr.16: 48084053-48084053 on Build GRCh38
rs75028443 Chr.16: 48084052-48084052 on Build GRCh38
rs765349844 Chr.16: 48083890-48083890 on Build GRCh38
rs766139127 Chr.16: 48083877-48083877 on Build GRCh38
rs746236687 Chr.16: 48084024-48084024 on Build GRCh38
rs775536359 Chr.16: 48084079-48084079 on Build GRCh38
rs144810262 Chr.16: 48084045-48084045 on Build GRCh38
rs374923473 Chr.16: 48084077-48084077 on Build GRCh38
rs201342647 Chr.16: 48083933-48083933 on Build GRCh38
rs147250788 Chr.16: 48083939-48083939 on Build GRCh38
rs755022479 Chr.16: 48083889-48083889 on Build GRCh38

View Full Product Details