Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 5173894-5174163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL8B
Entrez Gene ID
55207
Gene Name
ADP ribosylation factor like GTPase 8B
Gene Aliases
ARL10C, Gie1
UniGene
Hs.250009
Gene Chromosome Location
Chr.3: 5122245-5180916 on Build GRCh38
Transcripts
NM_018184.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6253714 c.414G>T p.L138F Chr.3: 5174058-5174058 on Build GRCh38
6468990 c.427C>T p.L143L Chr.3: 5174071-5174071 on Build GRCh38
6604857 c.434delA p.M147fs*1 Chr.3: 5174078-5174078 on Build GRCh38
6338317 c.394A>G p.R132G Chr.3: 5174038-5174038 on Build GRCh38
480269 c.397G>C p.D133H Chr.3: 5174041-5174041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77402822 Chr.3: 5174122-5174122 on Build GRCh38
rs1002650593 Chr.3: 5173981-5173981 on Build GRCh38
rs749746433 Chr.3: 5173986-5173986 on Build GRCh38
rs1012930806 Chr.3: 5173968-5173968 on Build GRCh38
rs750194211 Chr.3: 5174073-5174073 on Build GRCh38
rs1031503217 Chr.3: 5174101-5174101 on Build GRCh38
rs553989442 Chr.3: 5173987-5173987 on Build GRCh38
rs1037678234 Chr.3: 5173943-5173943 on Build GRCh38
rs531119865 Chr.3: 5173947-5173950 on Build GRCh38
rs756304631 Chr.3: 5174131-5174131 on Build GRCh38
rs534014535 Chr.3: 5173985-5173985 on Build GRCh38
rs776302140 Chr.3: 5174022-5174022 on Build GRCh38
rs761406964 Chr.3: 5174044-5174044 on Build GRCh38
rs765012161 Chr.3: 5174064-5174064 on Build GRCh38
rs766488279 Chr.3: 5174085-5174085 on Build GRCh38
rs888700766 Chr.3: 5173924-5173928 on Build GRCh38
rs866588237 Chr.3: 5173949-5173949 on Build GRCh38
rs748436773 Chr.3: 5173974-5173974 on Build GRCh38
rs567321573 Chr.3: 5174084-5174084 on Build GRCh38
rs9814309 Chr.3: 5173920-5173920 on Build GRCh38
rs760220235 Chr.3: 5174000-5174000 on Build GRCh38
rs940230380 Chr.3: 5173924-5173924 on Build GRCh38
rs1004338739 Chr.3: 5173947-5173947 on Build GRCh38
rs190144581 Chr.3: 5173955-5173955 on Build GRCh38
rs762615166 Chr.3: 5174035-5174036 on Build GRCh38
rs770146102 Chr.3: 5173976-5173976 on Build GRCh38
rs751657524 Chr.3: 5174095-5174095 on Build GRCh38
rs1007350844 Chr.3: 5174002-5174002 on Build GRCh38
rs781444170 Chr.3: 5174104-5174104 on Build GRCh38
rs1023278033 Chr.3: 5173970-5173970 on Build GRCh38
rs755155546 Chr.3: 5174103-5174103 on Build GRCh38
rs367665343 Chr.3: 5174019-5174019 on Build GRCh38

View Full Product Details