Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22182835-22183082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA6
Entrez Gene ID
2627
Gene Name
GATA binding protein 6
Gene Aliases
-
UniGene
Hs.514746
Gene Chromosome Location
Chr.18: 22169437-22202528 on Build GRCh38
Transcripts
NM_005257.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
237438 c.1516+4A>G p.? Chr.18: 22182848-22182848 on Build GRCh38
1750467 c.1616C>G p.S539* Chr.18: 22183039-22183039 on Build GRCh38
6669589 c.1602A>G p.T534T Chr.18: 22183025-22183025 on Build GRCh38
3726704 c.1620+6T>C p.? Chr.18: 22183049-22183049 on Build GRCh38
4071464 c.1507A>T p.T503S Chr.18: 22182835-22182835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200795241 Chr.18: 22183049-22183049 on Build GRCh38
rs769383865 Chr.18: 22182988-22182988 on Build GRCh38
rs113144042 Chr.18: 22182990-22182990 on Build GRCh38
rs774738165 Chr.18: 22182887-22182887 on Build GRCh38
rs1023324008 Chr.18: 22182960-22182960 on Build GRCh38
rs978211774 Chr.18: 22182982-22182982 on Build GRCh38
rs924066911 Chr.18: 22183015-22183015 on Build GRCh38
rs758195227 Chr.18: 22182916-22182916 on Build GRCh38
rs3764962 Chr.18: 22183050-22183050 on Build GRCh38
rs1025404539 Chr.18: 22182879-22182879 on Build GRCh38
rs373712868 Chr.18: 22182897-22182897 on Build GRCh38
rs780615005 Chr.18: 22182978-22182978 on Build GRCh38
rs762370751 Chr.18: 22182896-22182896 on Build GRCh38
rs769203773 Chr.18: 22183013-22183013 on Build GRCh38
rs766611274 Chr.18: 22182908-22182908 on Build GRCh38
rs774778659 Chr.18: 22183019-22183019 on Build GRCh38
rs369486263 Chr.18: 22182878-22182878 on Build GRCh38
rs748888875 Chr.18: 22183004-22183004 on Build GRCh38
rs369809143 Chr.18: 22183032-22183032 on Build GRCh38
rs954047630 Chr.18: 22182903-22182903 on Build GRCh38
rs140212942 Chr.18: 22182949-22182949 on Build GRCh38
rs374604452 Chr.18: 22182919-22182919 on Build GRCh38
rs117646477 Chr.18: 22183028-22183028 on Build GRCh38
rs754022225 Chr.18: 22182909-22182909 on Build GRCh38
rs746983527 Chr.18: 22182943-22182943 on Build GRCh38
rs199684979 Chr.18: 22182995-22182995 on Build GRCh38
rs202044652 Chr.18: 22182885-22182885 on Build GRCh38
rs956872746 Chr.18: 22183029-22183029 on Build GRCh38
rs750488881 Chr.18: 22182899-22182899 on Build GRCh38
rs569368270 Chr.18: 22182980-22182980 on Build GRCh38
rs756378515 Chr.18: 22182906-22182906 on Build GRCh38
rs146949786 Chr.18: 22183025-22183025 on Build GRCh38

View Full Product Details