Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44622507-44622780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADA
Entrez Gene ID
100
Gene Name
adenosine deaminase
Gene Aliases
-
UniGene
Hs.654536
Gene Chromosome Location
Chr.20: 44619519-44651758 on Build GRCh38
Transcripts
NM_000022.3 NM_001322050.1 NM_001322051.1 NR_136160.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748172 c.815G>A p.W272* Chr.20: 44622618-44622618 on Build GRCh38
4595808 c.802C>T p.L268F Chr.20: 44622631-44622631 on Build GRCh38
4595807 c.801C>A p.Y267* Chr.20: 44622632-44622632 on Build GRCh38
1027114 c.792G>A p.W264* Chr.20: 44622641-44622641 on Build GRCh38
6267514 c.813C>A p.A271A Chr.20: 44622620-44622620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559798694 Chr.20: 44622589-44622589 on Build GRCh38
rs764386125 Chr.20: 44622695-44622695 on Build GRCh38
rs771213761 Chr.20: 44622669-44622669 on Build GRCh38
rs528680964 Chr.20: 44622596-44622596 on Build GRCh38
rs759743522 Chr.20: 44622687-44622687 on Build GRCh38
rs773349681 Chr.20: 44622559-44622559 on Build GRCh38
rs994812351 Chr.20: 44622752-44622752 on Build GRCh38
rs748088317 Chr.20: 44622648-44622648 on Build GRCh38
rs899193277 Chr.20: 44622740-44622740 on Build GRCh38
rs374812597 Chr.20: 44622656-44622656 on Build GRCh38
rs757796081 Chr.20: 44622647-44622648 on Build GRCh38
rs570514588 Chr.20: 44622707-44622707 on Build GRCh38
rs746370947 Chr.20: 44622690-44622690 on Build GRCh38
rs370450947 Chr.20: 44622633-44622633 on Build GRCh38
rs550893200 Chr.20: 44622666-44622666 on Build GRCh38
rs754172068 Chr.20: 44622701-44622701 on Build GRCh38
rs934826988 Chr.20: 44622689-44622689 on Build GRCh38
rs116962828 Chr.20: 44622599-44622599 on Build GRCh38
rs774708098 Chr.20: 44622674-44622674 on Build GRCh38
rs542229902 Chr.20: 44622602-44622602 on Build GRCh38
rs577405975 Chr.20: 44622531-44622531 on Build GRCh38
rs1013028412 Chr.20: 44622723-44622723 on Build GRCh38
rs763198534 Chr.20: 44622571-44622571 on Build GRCh38
rs895973555 Chr.20: 44622719-44622719 on Build GRCh38
rs773016155 Chr.20: 44622664-44622664 on Build GRCh38
rs766590645 Chr.20: 44622587-44622587 on Build GRCh38
rs121908738 Chr.20: 44622612-44622612 on Build GRCh38
rs562095440 Chr.20: 44622611-44622611 on Build GRCh38
rs761204032 Chr.20: 44622692-44622692 on Build GRCh38
rs756480576 Chr.20: 44622606-44622606 on Build GRCh38
rs778167423 Chr.20: 44622608-44622608 on Build GRCh38
rs182482421 Chr.20: 44622753-44622753 on Build GRCh38
rs751635016 Chr.20: 44622588-44622588 on Build GRCh38
rs761572248 Chr.20: 44622554-44622554 on Build GRCh38
rs973595980 Chr.20: 44622534-44622534 on Build GRCh38
rs904398666 Chr.20: 44622705-44622705 on Build GRCh38
rs1057292007 Chr.20: 44622710-44622710 on Build GRCh38
rs781040248 Chr.20: 44622640-44622640 on Build GRCh38
rs767679026 Chr.20: 44622691-44622691 on Build GRCh38
rs1044495128 Chr.20: 44622738-44622738 on Build GRCh38
rs776548198 Chr.20: 44622550-44622550 on Build GRCh38
rs781605283 Chr.20: 44622655-44622656 on Build GRCh38
rs545935345 Chr.20: 44622558-44622558 on Build GRCh38
rs1000953688 Chr.20: 44622734-44622736 on Build GRCh38
rs760746989 Chr.20: 44622533-44622533 on Build GRCh38
rs769511813 Chr.20: 44622661-44622661 on Build GRCh38

View Full Product Details