Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 56623689-56623948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
UniGene
Hs.116877
Gene Chromosome Location
Chr.3: 56620132-56683242 on Build GRCh38
Transcripts
NM_001112736.1 NM_015224.3 XM_005264999.1 XM_006713077.2 XM_006713078.2 XM_011533552.2 XM_011533553.2 XM_017006030.1 XM_017006031.1 XM_017006032.1

SNP Detail

SNP ID Chromosome Location
rs998252721 Chr.3: 56623755-56623755 on Build GRCh38
rs976938290 Chr.3: 56623889-56623889 on Build GRCh38
rs751380219 Chr.3: 56623853-56623853 on Build GRCh38
rs965032658 Chr.3: 56623884-56623884 on Build GRCh38
rs1022977014 Chr.3: 56623888-56623889 on Build GRCh38
rs957108678 Chr.3: 56623855-56623855 on Build GRCh38
rs141000613 Chr.3: 56623871-56623871 on Build GRCh38
rs777503289 Chr.3: 56623772-56623772 on Build GRCh38
rs565787244 Chr.3: 56623883-56623887 on Build GRCh38
rs1018265667 Chr.3: 56623719-56623719 on Build GRCh38
rs957844280 Chr.3: 56623730-56623730 on Build GRCh38
rs561775495 Chr.3: 56623856-56623856 on Build GRCh38
rs910910316 Chr.3: 56623863-56623863 on Build GRCh38
rs879433730 Chr.3: 56623907-56623908 on Build GRCh38
rs1029755536 Chr.3: 56623796-56623796 on Build GRCh38
rs769588554 Chr.3: 56623805-56623806 on Build GRCh38
rs762236481 Chr.3: 56623908-56623908 on Build GRCh38
rs918849219 Chr.3: 56623907-56623907 on Build GRCh38

View Full Product Details