Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135395759-135395996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT4
Entrez Gene ID
4850
Gene Name
CCR4-NOT transcription complex subunit 4
Gene Aliases
CLONE243, NOT4, NOT4H
UniGene
Hs.490224
Gene Chromosome Location
Chr.7: 135361795-135510127 on Build GRCh38
Transcripts
NM_001008225.2 NM_001190847.1 NM_001190848.1 NM_001190849.1 NM_001190850.1 NM_013316.3 XM_017012235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2859098 c.953G>T p.S318I Chr.7: 135395801-135395801 on Build GRCh38
3633655 c.940C>T p.P314S Chr.7: 135395823-135395823 on Build GRCh38
1201610 c.968G>A p.R323Q Chr.7: 135395795-135395795 on Build GRCh38
7361736 c.926C>G p.S309C Chr.7: 135395837-135395837 on Build GRCh38
1085837 c.960G>T p.R320R Chr.7: 135395794-135395794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028342048 Chr.7: 135395941-135395941 on Build GRCh38
rs757342148 Chr.7: 135395849-135395849 on Build GRCh38
rs750612832 Chr.7: 135395860-135395860 on Build GRCh38
rs763252638 Chr.7: 135395821-135395821 on Build GRCh38
rs781312469 Chr.7: 135395855-135395855 on Build GRCh38
rs770474891 Chr.7: 135395892-135395892 on Build GRCh38
rs1008602068 Chr.7: 135395850-135395850 on Build GRCh38
rs756376765 Chr.7: 135395866-135395866 on Build GRCh38
rs771770362 Chr.7: 135395879-135395879 on Build GRCh38
rs771995836 Chr.7: 135395785-135395785 on Build GRCh38
rs564908475 Chr.7: 135395795-135395795 on Build GRCh38
rs199584772 Chr.7: 135395790-135395790 on Build GRCh38
rs561008472 Chr.7: 135395920-135395920 on Build GRCh38
rs762265604 Chr.7: 135395932-135395932 on Build GRCh38
rs759333609 Chr.7: 135395796-135395796 on Build GRCh38
rs546146099 Chr.7: 135395823-135395823 on Build GRCh38
rs201613355 Chr.7: 135395867-135395867 on Build GRCh38
rs759244046 Chr.7: 135395906-135395906 on Build GRCh38
rs746977444 Chr.7: 135395888-135395888 on Build GRCh38
rs371021495 Chr.7: 135395845-135395845 on Build GRCh38
rs763386068 Chr.7: 135395947-135395947 on Build GRCh38
rs369784414 Chr.7: 135395963-135395963 on Build GRCh38
rs902671432 Chr.7: 135395943-135395943 on Build GRCh38
rs773187583 Chr.7: 135395916-135395917 on Build GRCh38
rs765203005 Chr.7: 135395919-135395920 on Build GRCh38
rs376812510 Chr.7: 135395842-135395842 on Build GRCh38
rs769524818 Chr.7: 135395907-135395907 on Build GRCh38
rs773056576 Chr.7: 135395786-135395786 on Build GRCh38
rs759447810 Chr.7: 135395918-135395919 on Build GRCh38
rs777708358 Chr.7: 135395883-135395883 on Build GRCh38
rs1053872459 Chr.7: 135395952-135395952 on Build GRCh38
rs752488563 Chr.7: 135395920-135395921 on Build GRCh38
rs772153185 Chr.7: 135395904-135395905 on Build GRCh38
rs962002673 Chr.7: 135395871-135395871 on Build GRCh38
rs776325860 Chr.7: 135395905-135395905 on Build GRCh38
rs74772890 Chr.7: 135395813-135395813 on Build GRCh38
rs368236968 Chr.7: 135395914-135395914 on Build GRCh38
rs753163623 Chr.7: 135395818-135395818 on Build GRCh38
rs752099319 Chr.7: 135395924-135395924 on Build GRCh38
rs762797227 Chr.7: 135395921-135395922 on Build GRCh38

View Full Product Details