Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14457739-14457988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPB1
Entrez Gene ID
1315
Gene Name
coatomer protein complex subunit beta 1
Gene Aliases
COPB
UniGene
Hs.339278
Gene Chromosome Location
Chr.11: 14457503-14499895 on Build GRCh38
Transcripts
NM_001144061.1 NM_001144062.1 NM_016451.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1352822 c.2852C>T p.T951I Chr.11: 14457834-14457834 on Build GRCh38
3700023 c.2857A>G p.I953V Chr.11: 14457829-14457829 on Build GRCh38
6132029 c.2840C>T p.S947L Chr.11: 14457846-14457846 on Build GRCh38
4791701 c.2809G>A p.A937T Chr.11: 14457877-14457877 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372019218 Chr.11: 14457782-14457782 on Build GRCh38
rs775998909 Chr.11: 14457887-14457887 on Build GRCh38
rs752682987 Chr.11: 14457934-14457934 on Build GRCh38
rs746341161 Chr.11: 14457880-14457880 on Build GRCh38
rs757674503 Chr.11: 14457813-14457816 on Build GRCh38
rs768685954 Chr.11: 14457892-14457892 on Build GRCh38
rs532426996 Chr.11: 14457928-14457928 on Build GRCh38
rs766145525 Chr.11: 14457781-14457781 on Build GRCh38
rs755010104 Chr.11: 14457794-14457794 on Build GRCh38
rs771309231 Chr.11: 14457839-14457839 on Build GRCh38
rs770011031 Chr.11: 14457888-14457893 on Build GRCh38
rs749325380 Chr.11: 14457904-14457905 on Build GRCh38
rs781719255 Chr.11: 14457879-14457880 on Build GRCh38
rs775591113 Chr.11: 14457903-14457904 on Build GRCh38
rs777415338 Chr.11: 14457828-14457828 on Build GRCh38
rs761991539 Chr.11: 14457898-14457898 on Build GRCh38
rs915323244 Chr.11: 14457958-14457958 on Build GRCh38
rs988665796 Chr.11: 14457899-14457899 on Build GRCh38
rs754013176 Chr.11: 14457778-14457778 on Build GRCh38
rs372384010 Chr.11: 14457769-14457769 on Build GRCh38
rs755695147 Chr.11: 14457816-14457816 on Build GRCh38
rs760682268 Chr.11: 14457891-14457891 on Build GRCh38
rs200690876 Chr.11: 14457808-14457808 on Build GRCh38
rs374325633 Chr.11: 14457772-14457772 on Build GRCh38
rs768793573 Chr.11: 14457915-14457916 on Build GRCh38
rs779595842 Chr.11: 14457795-14457795 on Build GRCh38
rs776900167 Chr.11: 14457895-14457895 on Build GRCh38
rs761792234 Chr.11: 14457918-14457923 on Build GRCh38
rs542580626 Chr.11: 14457955-14457955 on Build GRCh38
rs781311582 Chr.11: 14457806-14457806 on Build GRCh38
rs371492671 Chr.11: 14457909-14457909 on Build GRCh38
rs758208885 Chr.11: 14457780-14457780 on Build GRCh38
rs774626201 Chr.11: 14457877-14457877 on Build GRCh38
rs748955941 Chr.11: 14457829-14457829 on Build GRCh38
rs746251442 Chr.11: 14457886-14457886 on Build GRCh38

View Full Product Details