Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219514212-219514485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC4
Entrez Gene ID
55515
Gene Name
acid sensing ion channel subunit family member 4
Gene Aliases
ACCN4, BNAC4
UniGene
Hs.87469
Gene Chromosome Location
Chr.2: 219514170-219538772 on Build GRCh38
Transcripts
NM_182847.2 XM_005246671.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6421896 c.128A>G p.K43R Chr.2: 219514471-219514471 on Build GRCh38
5682481 c.42A>T p.A14A Chr.2: 219514385-219514385 on Build GRCh38
1405670 c.65G>A p.R22H Chr.2: 219514408-219514408 on Build GRCh38
5527013 c.52C>T p.P18S Chr.2: 219514395-219514395 on Build GRCh38
1405671 c.122G>A p.S41N Chr.2: 219514465-219514465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761265425 Chr.2: 219514328-219514328 on Build GRCh38
rs766898377 Chr.2: 219514397-219514397 on Build GRCh38
rs949728516 Chr.2: 219514352-219514352 on Build GRCh38
rs775670225 Chr.2: 219514260-219514260 on Build GRCh38
rs758154207 Chr.2: 219514357-219514357 on Build GRCh38
rs762206552 Chr.2: 219514306-219514306 on Build GRCh38
rs1040332974 Chr.2: 219514317-219514317 on Build GRCh38
rs749708175 Chr.2: 219514269-219514269 on Build GRCh38
rs1018868210 Chr.2: 219514265-219514265 on Build GRCh38
rs772516020 Chr.2: 219514391-219514391 on Build GRCh38
rs752351441 Chr.2: 219514353-219514353 on Build GRCh38
rs773601560 Chr.2: 219514392-219514392 on Build GRCh38
rs115764695 Chr.2: 219514388-219514388 on Build GRCh38
rs990224014 Chr.2: 219514276-219514276 on Build GRCh38
rs1010035652 Chr.2: 219514380-219514380 on Build GRCh38
rs999936803 Chr.2: 219514371-219514371 on Build GRCh38
rs1053201914 Chr.2: 219514351-219514377 on Build GRCh38
rs771975031 Chr.2: 219514248-219514248 on Build GRCh38
rs375092746 Chr.2: 219514304-219514304 on Build GRCh38
rs532356410 Chr.2: 219514246-219514246 on Build GRCh38
rs774644784 Chr.2: 219514379-219514379 on Build GRCh38
rs984911705 Chr.2: 219514411-219514411 on Build GRCh38
rs768866608 Chr.2: 219514375-219514375 on Build GRCh38
rs992352134 Chr.2: 219514431-219514431 on Build GRCh38
rs966408351 Chr.2: 219514274-219514274 on Build GRCh38
rs931794066 Chr.2: 219514405-219514408 on Build GRCh38
rs753385420 Chr.2: 219514336-219514336 on Build GRCh38
rs764632026 Chr.2: 219514294-219514294 on Build GRCh38
rs952130026 Chr.2: 219514286-219514286 on Build GRCh38
rs774767733 Chr.2: 219514303-219514303 on Build GRCh38
rs61747688 Chr.2: 219514395-219514395 on Build GRCh38
rs759155962 Chr.2: 219514260-219514260 on Build GRCh38
rs773444387 Chr.2: 219514401-219514404 on Build GRCh38
rs542326823 Chr.2: 219514373-219514373 on Build GRCh38
rs576818700 Chr.2: 219514323-219514323 on Build GRCh38
rs923850073 Chr.2: 219514302-219514302 on Build GRCh38
rs369115588 Chr.2: 219514412-219514412 on Build GRCh38
rs908073896 Chr.2: 219514289-219514289 on Build GRCh38
rs545925064 Chr.2: 219514337-219514337 on Build GRCh38
rs371897118 Chr.2: 219514372-219514372 on Build GRCh38
rs559702545 Chr.2: 219514358-219514358 on Build GRCh38
rs962170477 Chr.2: 219514416-219514416 on Build GRCh38
rs562970540 Chr.2: 219514400-219514400 on Build GRCh38
rs369295055 Chr.2: 219514333-219514333 on Build GRCh38
rs766043612 Chr.2: 219514426-219514426 on Build GRCh38
rs940417683 Chr.2: 219514415-219514415 on Build GRCh38
rs779336608 Chr.2: 219514271-219514271 on Build GRCh38
rs764758273 Chr.2: 219514408-219514408 on Build GRCh38
rs752410085 Chr.2: 219514404-219514405 on Build GRCh38
rs562239101 Chr.2: 219514374-219514374 on Build GRCh38
rs1029022302 Chr.2: 219514399-219514399 on Build GRCh38
rs758027460 Chr.2: 219514448-219514448 on Build GRCh38
rs1021693013 Chr.2: 219514383-219514383 on Build GRCh38
rs918182324 Chr.2: 219514351-219514351 on Build GRCh38
rs543292891 Chr.2: 219514280-219514280 on Build GRCh38
rs556872394 Chr.2: 219514295-219514295 on Build GRCh38
rs767998239 Chr.2: 219514322-219514322 on Build GRCh38
rs115032706 Chr.2: 219514453-219514453 on Build GRCh38
rs574312074 Chr.2: 219514279-219514279 on Build GRCh38
rs761121744 Chr.2: 219514288-219514288 on Build GRCh38
rs1032837336 Chr.2: 219514237-219514237 on Build GRCh38
rs779433368 Chr.2: 219514407-219514407 on Build GRCh38
rs528539886 Chr.2: 219514370-219514370 on Build GRCh38

View Full Product Details