Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75914668-75914941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBF1
Entrez Gene ID
85302
Gene Name
Fas binding factor 1
Gene Aliases
Alb, FBF-1
UniGene
Hs.720068
Gene Chromosome Location
Chr.17: 75909574-75938232 on Build GRCh38
Transcripts
NM_001319193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6661460 c.2661G>A p.A887A Chr.17: 75914855-75914855 on Build GRCh38
4069722 c.2642G>A p.R881H Chr.17: 75914874-75914874 on Build GRCh38
4590459 c.2708A>G p.E903G Chr.17: 75914808-75914808 on Build GRCh38
1195096 c.2641C>T p.R881C Chr.17: 75914875-75914875 on Build GRCh38
1480024 c.2707G>A p.E903K Chr.17: 75914809-75914809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751411820 Chr.17: 75914791-75914791 on Build GRCh38
rs562007185 Chr.17: 75914843-75914843 on Build GRCh38
rs369687550 Chr.17: 75914889-75914889 on Build GRCh38
rs749865465 Chr.17: 75914776-75914776 on Build GRCh38
rs754147611 Chr.17: 75914805-75914805 on Build GRCh38
rs560252459 Chr.17: 75914747-75914747 on Build GRCh38
rs535838759 Chr.17: 75914846-75914846 on Build GRCh38
rs573986983 Chr.17: 75914803-75914803 on Build GRCh38
rs779091042 Chr.17: 75914809-75914809 on Build GRCh38
rs186970426 Chr.17: 75914851-75914851 on Build GRCh38
rs761370334 Chr.17: 75914771-75914771 on Build GRCh38
rs748408284 Chr.17: 75914903-75914903 on Build GRCh38
rs753547006 Chr.17: 75914796-75914816 on Build GRCh38
rs376953918 Chr.17: 75914902-75914902 on Build GRCh38
rs577743025 Chr.17: 75914699-75914699 on Build GRCh38
rs757398787 Chr.17: 75914808-75914808 on Build GRCh38
rs577460839 Chr.17: 75914799-75914810 on Build GRCh38
rs550825443 Chr.17: 75914855-75914855 on Build GRCh38
rs755999915 Chr.17: 75914705-75914716 on Build GRCh38
rs772428036 Chr.17: 75914733-75914733 on Build GRCh38
rs758408470 Chr.17: 75914895-75914895 on Build GRCh38
rs368877297 Chr.17: 75914775-75914775 on Build GRCh38
rs775866087 Chr.17: 75914736-75914736 on Build GRCh38
rs907231532 Chr.17: 75914863-75914863 on Build GRCh38
rs780106055 Chr.17: 75914896-75914896 on Build GRCh38
rs766368547 Chr.17: 75914789-75914789 on Build GRCh38
rs781129491 Chr.17: 75914804-75914804 on Build GRCh38
rs750531702 Chr.17: 75914891-75914891 on Build GRCh38
rs777674249 Chr.17: 75914723-75914728 on Build GRCh38
rs755459098 Chr.17: 75914900-75914900 on Build GRCh38
rs374783310 Chr.17: 75914767-75914767 on Build GRCh38
rs917805879 Chr.17: 75914723-75914723 on Build GRCh38
rs200319918 Chr.17: 75914890-75914890 on Build GRCh38
rs541709568 Chr.17: 75914816-75914816 on Build GRCh38
rs769497260 Chr.17: 75914904-75914904 on Build GRCh38
rs564296983 Chr.17: 75914865-75914865 on Build GRCh38
rs369144467 Chr.17: 75914907-75914907 on Build GRCh38
rs1038383863 Chr.17: 75914820-75914820 on Build GRCh38
rs1042385027 Chr.17: 75914729-75914729 on Build GRCh38
rs747575863 Chr.17: 75914897-75914897 on Build GRCh38
rs201707040 Chr.17: 75914888-75914888 on Build GRCh38
rs769079564 Chr.17: 75914840-75914840 on Build GRCh38
rs941533222 Chr.17: 75914813-75914813 on Build GRCh38
rs776202390 Chr.17: 75914756-75914756 on Build GRCh38
rs889882623 Chr.17: 75914811-75914811 on Build GRCh38
rs994858630 Chr.17: 75914868-75914868 on Build GRCh38
rs1025965803 Chr.17: 75914878-75914878 on Build GRCh38
rs748964911 Chr.17: 75914920-75914920 on Build GRCh38
rs760980698 Chr.17: 75914746-75914746 on Build GRCh38
rs762976258 Chr.17: 75914784-75914784 on Build GRCh38
rs780483273 Chr.17: 75914822-75914822 on Build GRCh38
rs772798458 Chr.17: 75914861-75914861 on Build GRCh38
rs766421890 Chr.17: 75914872-75914872 on Build GRCh38
rs372404725 Chr.17: 75914794-75914794 on Build GRCh38
rs964796339 Chr.17: 75914915-75914915 on Build GRCh38
rs745896743 Chr.17: 75914810-75914810 on Build GRCh38
rs751612554 Chr.17: 75914879-75914879 on Build GRCh38
rs371937178 Chr.17: 75914710-75914710 on Build GRCh38
rs202117712 Chr.17: 75914728-75914728 on Build GRCh38
rs925305240 Chr.17: 75914769-75914769 on Build GRCh38
rs372207421 Chr.17: 75914833-75914833 on Build GRCh38
rs999150652 Chr.17: 75914873-75914873 on Build GRCh38
rs936752445 Chr.17: 75914785-75914785 on Build GRCh38
rs756877254 Chr.17: 75914797-75914797 on Build GRCh38

View Full Product Details