Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22171253-22171478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA6
Entrez Gene ID
2627
Gene Name
GATA binding protein 6
Gene Aliases
-
UniGene
Hs.514746
Gene Chromosome Location
Chr.18: 22169437-22202528 on Build GRCh38
Transcripts
NM_005257.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3773369 c.238C>T p.L80F Chr.18: 22171382-22171382 on Build GRCh38
5694144 c.331G>T p.D111Y Chr.18: 22171475-22171475 on Build GRCh38
6886123 c.228C>T p.R76R Chr.18: 22171372-22171372 on Build GRCh38
3388389 c.134C>G p.S45C Chr.18: 22171278-22171278 on Build GRCh38
6217963 c.209C>T p.A70V Chr.18: 22171353-22171353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749200220 Chr.18: 22171451-22171451 on Build GRCh38
rs750899923 Chr.18: 22171326-22171326 on Build GRCh38
rs374402554 Chr.18: 22171366-22171366 on Build GRCh38
rs758629279 Chr.18: 22171440-22171440 on Build GRCh38
rs752927641 Chr.18: 22171432-22171432 on Build GRCh38
rs377217497 Chr.18: 22171327-22171327 on Build GRCh38
rs767886137 Chr.18: 22171318-22171318 on Build GRCh38
rs924843423 Chr.18: 22171316-22171316 on Build GRCh38
rs755180659 Chr.18: 22171286-22171286 on Build GRCh38
rs762294271 Chr.18: 22171315-22171315 on Build GRCh38
rs200483324 Chr.18: 22171347-22171347 on Build GRCh38
rs772338637 Chr.18: 22171381-22171381 on Build GRCh38
rs756058812 Chr.18: 22171448-22171448 on Build GRCh38
rs554577705 Chr.18: 22171340-22171340 on Build GRCh38
rs751845516 Chr.18: 22171409-22171409 on Build GRCh38
rs773613709 Chr.18: 22171394-22171394 on Build GRCh38
rs923229630 Chr.18: 22171343-22171343 on Build GRCh38
rs746782842 Chr.18: 22171292-22171292 on Build GRCh38
rs993070434 Chr.18: 22171361-22171361 on Build GRCh38
rs750205839 Chr.18: 22171445-22171445 on Build GRCh38
rs770662152 Chr.18: 22171295-22171295 on Build GRCh38
rs762765948 Chr.18: 22171313-22171313 on Build GRCh38
rs759641995 Chr.18: 22171418-22171418 on Build GRCh38
rs767279089 Chr.18: 22171444-22171444 on Build GRCh38
rs529881815 Chr.18: 22171421-22171421 on Build GRCh38
rs778509273 Chr.18: 22171454-22171454 on Build GRCh38
rs780979305 Chr.18: 22171345-22171345 on Build GRCh38
rs563978220 Chr.18: 22171403-22171403 on Build GRCh38
rs978912698 Chr.18: 22171308-22171308 on Build GRCh38
rs200519946 Chr.18: 22171297-22171297 on Build GRCh38
rs374591296 Chr.18: 22171426-22171426 on Build GRCh38
rs761329606 Chr.18: 22171293-22171293 on Build GRCh38
rs941842417 Chr.18: 22171414-22171415 on Build GRCh38
rs767885101 Chr.18: 22171280-22171280 on Build GRCh38
rs768396712 Chr.18: 22171314-22171314 on Build GRCh38
rs933199729 Chr.18: 22171344-22171344 on Build GRCh38
rs968804836 Chr.18: 22171272-22171272 on Build GRCh38
rs1044783206 Chr.18: 22171395-22171395 on Build GRCh38
rs780116515 Chr.18: 22171283-22171283 on Build GRCh38
rs773974211 Chr.18: 22171391-22171396 on Build GRCh38
rs541150137 Chr.18: 22171299-22171299 on Build GRCh38
rs762445989 Chr.18: 22171273-22171273 on Build GRCh38
rs747737834 Chr.18: 22171371-22171371 on Build GRCh38
rs765466198 Chr.18: 22171338-22171338 on Build GRCh38
rs779755261 Chr.18: 22171352-22171352 on Build GRCh38
rs745748080 Chr.18: 22171302-22171302 on Build GRCh38
rs780166588 Chr.18: 22171450-22171450 on Build GRCh38
rs766886560 Chr.18: 22171415-22171415 on Build GRCh38
rs777072362 Chr.18: 22171417-22171417 on Build GRCh38
rs748788731 Chr.18: 22171365-22171365 on Build GRCh38
rs766427554 Chr.18: 22171330-22171330 on Build GRCh38
rs774180483 Chr.18: 22171368-22171368 on Build GRCh38
rs756181712 Chr.18: 22171281-22171281 on Build GRCh38
rs754073585 Chr.18: 22171285-22171285 on Build GRCh38
rs559705145 Chr.18: 22171331-22171331 on Build GRCh38
rs867701103 Chr.18: 22171358-22171358 on Build GRCh38
rs777584552 Chr.18: 22171291-22171291 on Build GRCh38
rs769197296 Chr.18: 22171304-22171304 on Build GRCh38
rs36041440 Chr.18: 22171329-22171330 on Build GRCh38

View Full Product Details