Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45677835-45678084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY1
Entrez Gene ID
107
Gene Name
adenylate cyclase 1
Gene Aliases
AC1, DFNB44
UniGene
Hs.192215
Gene Chromosome Location
Chr.7: 45574140-45723116 on Build GRCh38
Transcripts
NM_021116.2 XM_005249584.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3881332 c.1660A>G p.T554A Chr.7: 45677923-45677923 on Build GRCh38
6843385 c.1687A>T p.T563S Chr.7: 45677950-45677950 on Build GRCh38
5679834 c.1606C>A p.R536R Chr.7: 45677869-45677869 on Build GRCh38
1698655 c.1735G>A p.E579K Chr.7: 45677998-45677998 on Build GRCh38
4928133 c.1717T>A p.L573I Chr.7: 45677980-45677980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759674723 Chr.7: 45677989-45677989 on Build GRCh38
rs760827696 Chr.7: 45678012-45678012 on Build GRCh38
rs370138365 Chr.7: 45677892-45677892 on Build GRCh38
rs780863849 Chr.7: 45678044-45678044 on Build GRCh38
rs762914964 Chr.7: 45677956-45677956 on Build GRCh38
rs751109490 Chr.7: 45678039-45678039 on Build GRCh38
rs367735209 Chr.7: 45677955-45677955 on Build GRCh38
rs147187783 Chr.7: 45678049-45678049 on Build GRCh38
rs754471725 Chr.7: 45678041-45678041 on Build GRCh38
rs142005939 Chr.7: 45678052-45678052 on Build GRCh38
rs370915655 Chr.7: 45678053-45678053 on Build GRCh38
rs781058119 Chr.7: 45677929-45677929 on Build GRCh38
rs754319377 Chr.7: 45677874-45677874 on Build GRCh38
rs371609367 Chr.7: 45678029-45678029 on Build GRCh38
rs140648069 Chr.7: 45678057-45678057 on Build GRCh38
rs770822019 Chr.7: 45677962-45677962 on Build GRCh38
rs201746307 Chr.7: 45677865-45677865 on Build GRCh38
rs778010377 Chr.7: 45677870-45677870 on Build GRCh38
rs747756051 Chr.7: 45678048-45678048 on Build GRCh38
rs757763607 Chr.7: 45678030-45678030 on Build GRCh38
rs35864133 Chr.7: 45677952-45677953 on Build GRCh38
rs767043187 Chr.7: 45677980-45677980 on Build GRCh38
rs768415059 Chr.7: 45677909-45677909 on Build GRCh38
rs143601874 Chr.7: 45677869-45677869 on Build GRCh38
rs779350746 Chr.7: 45678038-45678038 on Build GRCh38
rs75922090 Chr.7: 45677905-45677905 on Build GRCh38
rs747808945 Chr.7: 45677938-45677938 on Build GRCh38
rs746570916 Chr.7: 45677908-45677908 on Build GRCh38
rs764481666 Chr.7: 45678017-45678017 on Build GRCh38
rs774400678 Chr.7: 45677978-45677978 on Build GRCh38
rs752856161 Chr.7: 45678008-45678008 on Build GRCh38
rs767584608 Chr.7: 45677995-45677995 on Build GRCh38
rs779587943 Chr.7: 45677902-45677902 on Build GRCh38

View Full Product Details