Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88011026-88011279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR65
Entrez Gene ID
8477
Gene Name
G protein-coupled receptor 65
Gene Aliases
TDAG8, hTDAG8
UniGene
Hs.443243
Gene Chromosome Location
Chr.14: 88005124-88014811 on Build GRCh38
Transcripts
NM_003608.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
305621 c.326C>T p.A109V Chr.14: 88011173-88011173 on Build GRCh38
368056 c.432C>T p.I144I Chr.14: 88011279-88011279 on Build GRCh38
3728440 c.305C>T p.T102I Chr.14: 88011152-88011152 on Build GRCh38
385656 c.328G>T p.V110F Chr.14: 88011175-88011175 on Build GRCh38
1640165 c.364delT p.F124fs*2 Chr.14: 88011211-88011211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750989848 Chr.14: 88011210-88011210 on Build GRCh38
rs755830314 Chr.14: 88011219-88011219 on Build GRCh38
rs760532795 Chr.14: 88011190-88011190 on Build GRCh38
rs148741118 Chr.14: 88011099-88011099 on Build GRCh38
rs201126670 Chr.14: 88011128-88011128 on Build GRCh38
rs147419350 Chr.14: 88011191-88011191 on Build GRCh38
rs770829884 Chr.14: 88011211-88011211 on Build GRCh38
rs370595097 Chr.14: 88011199-88011199 on Build GRCh38
rs931847819 Chr.14: 88011127-88011127 on Build GRCh38
rs949144996 Chr.14: 88011245-88011245 on Build GRCh38
rs756852546 Chr.14: 88011133-88011133 on Build GRCh38
rs780853135 Chr.14: 88011148-88011148 on Build GRCh38
rs934967805 Chr.14: 88011215-88011215 on Build GRCh38
rs769208743 Chr.14: 88011174-88011174 on Build GRCh38
rs375417424 Chr.14: 88011212-88011212 on Build GRCh38
rs1055466 Chr.14: 88011220-88011220 on Build GRCh38
rs867394370 Chr.14: 88011202-88011202 on Build GRCh38
rs777146067 Chr.14: 88011100-88011100 on Build GRCh38
rs772388191 Chr.14: 88011182-88011182 on Build GRCh38
rs925220030 Chr.14: 88011132-88011132 on Build GRCh38
rs781204922 Chr.14: 88011090-88011090 on Build GRCh38
rs754855859 Chr.14: 88011058-88011058 on Build GRCh38
rs770118299 Chr.14: 88011159-88011159 on Build GRCh38
rs745560553 Chr.14: 88011158-88011158 on Build GRCh38
rs749751887 Chr.14: 88011172-88011172 on Build GRCh38
rs1055465 Chr.14: 88011138-88011138 on Build GRCh38
rs761978597 Chr.14: 88011181-88011181 on Build GRCh38
rs752555068 Chr.14: 88011097-88011097 on Build GRCh38
rs780214420 Chr.14: 88011230-88011230 on Build GRCh38
rs200487695 Chr.14: 88011233-88011233 on Build GRCh38
rs764112131 Chr.14: 88011201-88011201 on Build GRCh38
rs548128364 Chr.14: 88011175-88011175 on Build GRCh38
rs1050983323 Chr.14: 88011227-88011227 on Build GRCh38
rs752606649 Chr.14: 88011196-88011196 on Build GRCh38
rs1055467 Chr.14: 88011235-88011235 on Build GRCh38
rs566099533 Chr.14: 88011218-88011218 on Build GRCh38
rs748264608 Chr.14: 88011123-88011127 on Build GRCh38
rs772260251 Chr.14: 88011248-88011248 on Build GRCh38
rs377442975 Chr.14: 88011185-88011185 on Build GRCh38
rs6574978 Chr.14: 88011069-88011069 on Build GRCh38

View Full Product Details